SAB1405973

Sigma-Aldrich

Anti-HSPD1 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonim(y):
HSP60, HLD4, CPN60, HSP65, SPG13, HuCHA60, GROEL
NACRES:
NA.41

Poziom jakości

100

pochodzenie biologiczne

mouse

forma przeciwciała

purified immunoglobulin

antibody product type

primary antibodies

klon

polyclonal

formularz

buffered aqueous solution

masa cząsteczkowa

antigen ~61.1 kDa

species reactivity

human

zastosowania

indirect immunofluorescence: suitable
western blot: 1 μg/mL

białko sprzężone

unconjugated

numer dostępu NCBI

numer dostępu UniProt

wysyłka w ciągu

dry ice

temp. przechowywania

−20°C

Gene Information

human ... HSPD1(3329)

Opis ogólny

This gene encodes a member of the chaperonin family. The encoded mitochondrial protein may function as a signaling molecule in the innate immune system. This protein is essential for the folding and assembly of newly imported proteins in the mitochondria. This gene is adjacent to a related family member and the region between the 2 genes functions as a bidirectional promoter. Two pseudogenes, both located on chromosome 8, have been associated with this gene. Two transcript variants encoding the same protein have been identified for this gene. Mutations associated with this gene cause autosomal recessive spastic paraplegia 13. (provided by RefSeq)

Immunogen

HSPD1 (NP_002147.2, 1 a.a. ~ 573 a.a) full-length human protein.

Sequence
MLRLPTVFRQMRPVSRVLAPHLTRAYAKDVKFGADARALMLQGVDLLADAVAVTMGPKGRTVIIEQSWGSPKVTKDGVTVAKSIDLKDKYKNIGAKLVQDVANNTNEEAGDGTTTATVLARSIAKEGFEKISKGANPVEIRRGVMLAVDAVIAELKKQSKPVTTPEEIAQVATISANGDKEIGNIISDAMKKVGRKGVITVKDGKTLNDELEIIEGMKFDRGYISPYFINTSKGQKCEFQDAYVLLSEKKISSIQSIVPALEIANAHRKPLVIIAEDVDGEALSTLVLNRLKVGLQVVAVKAPGFGDNRKNQLKDMAIATGGAVFGEEGLTLNLEDVQPHDLGKVGEVIVTKDDAMLLKGKGDKAQIEKRIQEIIEQLDVTTSEYEKEKLNERLAKLSDGVAVLKVGGTSDVEVNEKKDRVTDALNATRAAVEEGIVLGGGCALLRCIPALDSLTPANEDQKIGIEIIKRTLKIPAMTIAKNAGVEGSLIVEKIMQSSSEVGYDAMAGDFVNMVEKGIIDPTKVVRTALLDAAGVASLLTTAEVVVTEIPKEEKDPGMGAMGGMGGGMGGGMF

Postać fizyczna

Solution in phosphate buffered saline, pH 7.4

Oświadczenie o zrzeczeniu się odpowiedzialności

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

storage_class_code

13 - Non Combustible Solids

WGK Germany

WGK 1

Temperatura zapłonu °F

Not applicable

Temperatura zapłonu °C

Not applicable

Certyfikat analizy

Świadectwo pochodzenia

Xiao-shan Li et al.
PloS one, 9(9), e107507-e107507 (2014-09-11)
Heat shock protein 60 (HSP60) is a chaperonin with essential functions for cell physiology and survival, and its expression correlates with prognosis in a number of malignancies. The aim of this study is to determine the relationship of HSP60 status...
Sylvie Bannwarth et al.
Brain : a journal of neurology, 137(Pt 8), 2329-2345 (2014-06-18)
Mitochondrial DNA instability disorders are responsible for a large clinical spectrum, among which amyotrophic lateral sclerosis-like symptoms and frontotemporal dementia are extremely rare. We report a large family with a late-onset phenotype including motor neuron disease, cognitive decline resembling frontotemporal...

Nasz zespół naukowców ma doświadczenie we wszystkich obszarach badań, w tym w naukach przyrodniczych, materiałoznawstwie, syntezie chemicznej, chromatografii, analityce i wielu innych dziedzinach.

Skontaktuj się z zespołem ds. pomocy technicznej