Wszystkie zdjęcia(1)

SAB4501178

Sigma-Aldrich

Anti-GJA3 antibody produced in rabbit

affinity isolated antibody

Synonim(y):
Connexin-46, Cx46, Gap junction α-3 protein
NACRES:
NA.41

pochodzenie biologiczne

rabbit

Poziom jakości

100

białko sprzężone

unconjugated

forma przeciwciała

affinity isolated antibody

antibody product type

primary antibodies

klon

polyclonal

forma

buffered aqueous solution

masa cząsteczkowa

antigen 47 kDa

species reactivity

mouse, human, rat

stężenie

~1 mg/mL

technique(s)

ELISA: 1:20000
immunohistochemistry: 1:50-1:100

numer dostępu NCBI

transport

wet ice

temp. przechowywania

−20°C

Gene Information

human ... CXA3(2700)

Opis ogólny

Anti-GJA3 Antibody detects endogenous levels of total GJA3 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human GJA3.

Immunogen Range: 151-200

Cechy i korzyści

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Postać fizyczna

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Kod klasy składowania

12 - Non Combustible Liquids

WGK

nwg

Temperatura zapłonu °F

Not applicable

Temperatura zapłonu °C

Not applicable

Certyfikat analizy

Świadectwo pochodzenia

Chang-Qing Fan et al.
International heart journal, 55(4), 362-371 (2014-06-27)
Impact of early bone marrow-derived mesenchymal stem cell (BMDMSC) implantation on left ventricular (LV) function after AMI was studied.Twelve mini-pigs were equally divided into placebo (AMI through left coronary artery ligation) and cell-treated groups [BMDMSCs (3.0 × 10(7)) implanted into
Appledene Osbourne et al.
Journal of molecular and cellular cardiology, 74, 53-63 (2014-05-14)
MicroRNAs (miRNAs) are now recognized as critical regulators of diverse physiological and pathological processes; however, studies of miRNAs and arrhythmogenesis remain sparse. Connexin43 (Cx43), a major cardiac gap junction protein, has elicited great interest in its role in arrhythmias. Additionally
Cindy X Guo et al.
Molecular vision, 20, 670-682 (2014-06-03)
Changes in connexin expression are associated with many pathological conditions seen in animal models and in humans. We hypothesized that gap junctions are important mediators in tissue dysfunction and injury processes in the retina, and therefore, we investigated the pattern
Daniel Soetkamp et al.
Basic research in cardiology, 109(5), 433-433 (2014-08-15)
S-nitrosation (SNO) of connexin 43 (Cx43)-formed channels modifies dye uptake in astrocytes and gap junctional communication in endothelial cells. Apart from forming channels at the plasma membrane of several cell types, Cx43 is also located at the inner membrane of

Nasz zespół naukowców ma doświadczenie we wszystkich obszarach badań, w tym w naukach przyrodniczych, materiałoznawstwie, syntezie chemicznej, chromatografii, analityce i wielu innych dziedzinach.

Skontaktuj się z zespołem ds. pomocy technicznej