Nazwa produktu
DL-Norleucine
Numer produktu
N1398
Marka produktu
SIGMA
Numer CAS
616-06-8
Masa cząsteczkowa
131.17
Test
Charakterystyka techniczna
Appearance (Color)White to Off-White
Appearance (Form)Powder
Solubility (Color)Colorless to Very Faint Yellow
Solubility (Turbidity)Clear to Slightly Hazy
 50 mg/ml in Formic Acid : H2O ( 1 : 4 )
Infrared spectrumConforms to Structure
Carbon53.8 - 56.0 %
Nitrogen10.4 - 11.0 %
Purity (TLC)≥98 %
Specific Rotation-0.5 - 0.5 °
 C = 1 in 5M HCl at 25 deg C