Nazwa produktu
Dowex 50W X8 hydrogen form, strongly acidic, 200-400 mesh
Numer produktu
44519
Marka produktu
sial
Numer CAS
69011-20-7
Masa cząsteczkowa
Test | Charakterystyka techniczna |
|---|---|
| APPEARANCE (COLOR) | White to Light Yellow to Light Beige |
| APPEARANCE (FORM) | Beads |
| REMARKS ON ANALYSIS | MAY CONTAIN BLACK PARTICLES |
| PARTICLE SIZE | CORRESPONDS TO REQUIREMENTS |
| IONIC TYPE | CORRESPONDS TO THE IONIC FORM |
| LOSS ON DRYING | 50 - 60 % |