Nazwa produktu
Anti-Bcl-2 (ab1) antibody produced in rabbit affinity isolated antibody, buffered aqueous solution
Numer produktu
prs3335
Marka produktu
sigma
Numer CAS
Masa cząsteczkowa
Test | Charakterystyka techniczna |
|---|---|
| Appearance | Report Result |
| Concentration | Report Result |
| Western Blotting | Bcl-2 antibody can be used for detection of Bcl-2 by Western blot at 1-2 µg/ml. |
| ELISA | Bcl-2 antibody can be used for detection of 10 ng of immunogenic Bcl-2 peptide by ELISA at 1 µg/ml. |
| Immunohistochemistry | Bcl-2 antibody can be used for immunohistochemistry at 2 µg/ml. |
| Immunofluorescence | Bcl-2 antibody can be used for immunofluorescence starting at 10 µg/ml. |