  • Search Results
  • Normal role of the low-molecular-weight neurofilament protein in mitochondrial dynamics and disruption in Charcot-Marie-Tooth disease.
Normal role of the low-molecular-weight neurofilament protein in mitochondrial dynamics and disruption in Charcot-Marie-Tooth disease.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2011-12-14)
Benoit J Gentil, Sandra Minotti, Madeleine Beange, Robert H Baloh, Jean-Pierre Julien, Heather D Durham
ABSTRACT
Intermediate filaments serve important structural roles, but other cellular functions are increasingly recognized. This study demonstrated normal function of the low-molecular-weight neurofilament protein (NFL) in mitochondrial dynamics and disruption in Charcot-Marie-Tooth disease (CMT) due to mutations in the Nefl gene. In motor neurons of spinal cord cultured from Nefl-knockout mice, mitochondrial length and the rate of fusion were decreased concomitant with increased motility. These parameters were normalized after expression of NFL(wt) on the Nefl(-/-) background, but not by overexpression of the profusion protein, mitofusin 2 (MFN2). The effects of CMT-causing NFL mutants bore similarities to and differences from Nefl knockout. In the early phase of toxicity before disruption of the neurofilament network, NFL(Q333P) and NFL(P8R) integrated into neurofilaments and had effects on mitochondria similar to those with Nefl knockout. The reduction of fusion rate by NFL(Q333P) was partly due to interference with the function of the profusion protein MFN2, which is mutated in CMT2A, functionally linking these forms of CMT. In the later phase of toxicity, mitochondria essentially stopped moving in neurons expressing NFL mutants, probably a consequence of cytoskeletal disruption. Overall, the data point to important functions of neurofilaments in mitochondrial dynamics as well as primary involvement in CMT2E/1F.
Sigma-Aldrich
Anti-Mitofusin-2 (N-Terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Neurofilament L Antibody, serum, Chemicon®