KANSL2 and MBNL3 are regulators of pancreatic ductal adenocarcinoma invasion.
Scientific reports (2020-02-01)
Peter O Oladimeji, Jesse Bakke, William C Wright, Taosheng Chen
ABSTRAKT
Pancreatic ductal adenocarcinoma (PDAC) is one of the most lethal forms of cancer. One major reason for this is that PDAC quickly metastasizes to other organs, thereby making its treatment difficult. The molecular machinery driving PDAC metastasis is still poorly understood. In this study, we applied an unbiased approach using CRISPR screening to identify genes that strongly regulate invasion (based on an in vitro assessment of their metastatic potential) in PANC-1, a PDAC cell line. Through CRISPR screening, we identified MBNL3 and KANSL2 as strong regulators of invasion in PANC-1 cells. We further validated MBNL3 and KANSL2 as regulators of PANC-1 cell invasion by using the doxycycline-inducible shRNA system. We also showed that MBNL3 and KANSL2 do not affect cell proliferation. Through our efforts, we have established a process to identify genes that regulate cell invasion and can be further investigated as potential targets for therapeutic intervention.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Puromycin dihydrochloride, Ready Made Solution, from Streptomyces alboniger, 10 mg/mL in H2O
Sigma-Aldrich
Hexadimethrine bromide, ≥95%