  • Search Results
  • Irreversible inactivation of trypanothione reductase by unsaturated Mannich bases: a divinyl ketone as key intermediate.
Irreversible inactivation of trypanothione reductase by unsaturated Mannich bases: a divinyl ketone as key intermediate.
Journal of medicinal chemistry (2005-11-11)
Brittany Lee, Holger Bauer, Johannes Melchers, Thomas Ruppert, Lauren Rattray, Vanessa Yardley, Elisabeth Davioud-Charvet, R Luise Krauth-Siegel
ABSTRACT
Trypanothione reductase is a flavoenzyme unique to trypanosomatid parasites. Here we show that unsaturated Mannich bases irreversibly inactivate trypanothione reductase from Trypanosoma cruzi, the causative agent of Chagas' disease. The inhibitory potency of the compounds strongly increased upon storage of the DMSO stock solutions. HPLC, NMR, and mass spectrometry data of potential intermediates revealed a divinyl ketone as the active compound inactivating the enzyme. ESI- and MALDI-TOF mass spectrometry of trypanothione reductase modified by the Mannich base or the divinyl ketone showed specific alkylation of the active site Cys52 by a 5-(2'chlorophenyl)-3-oxo-4-pentenyl substituent. The reaction mechanism and the site of alkylation differ from those in Plasmodium falciparum thioredoxin reductase where the C-terminal redox active dithiol is modified. After deamination, unsaturated Mannich bases are highly reactive in polycondensation with trypanothione. Interaction of these compounds with both trypanothione and trypanothione reductase could account for their potent trypanocidal effect against Trypanosoma brucei.
Sigma-Aldrich
Podophyllotoxin