  • Search Results
  • Synthesis and pharmacological evaluation of fluorescent and photoactivatable analogues of antiplasmodial naphthylisoquinolines.
Synthesis and pharmacological evaluation of fluorescent and photoactivatable analogues of antiplasmodial naphthylisoquinolines.
Journal of medicinal chemistry (2007-11-10)
Gerhard Bringmann, Christian M Gampe, Yanina Reichert, Torsten Bruhn, Johan H Faber, Martin Mikyna, Matthias Reichert, Matthias Leippe, Reto Brun, Christoph Gelhaus
ABSTRACT
The naphthylisoquinoline (NIQ) alkaloids from tropical Ancistrocladaceae and Dioncophyllaceae plants show high antiplasmodial activities in vitro and in vivo, even against chloroquine-resistant strains of the malaria pathogen. For the directed optimization of these activities, an investigation of the mode of action seems most rewarding. We have therefore embarked on the identification of the respective target protein in Plasmodium falciparum. For this purpose, we have developed a flexible pathway for the synthesis of a chemically divergent series of photoactive and fluorescent derivatives of such alkaloids and succeeded in preparing the first functionalized NIQ derivatives, 10, 12, and 35, suited for fluorescence and photoaffinity labeling experiments. Pharmacological investigations ensured that the modified alkaloid derivatives retained their antiplasmodial activity. The work may pave the way for a further improvement of the activity of these natural products and will thus increase their pharmacological potential as a valuable lead structure against the widespread tropical disease malaria.
Sigma-Aldrich
Podophyllotoxin