The DNA damage response links human squamous proliferation with differentiation.
The Journal of cell biology (2020-10-03)
Rut Molinuevo, Ana Freije, Lizbeth Contreras, Juan R Sanz, Alberto Gandarillas
ABSTRAKT
How rapid cell multiplication leads to cell differentiation in developing tissues is still enigmatic. This question is central to morphogenesis, cell number control, and homeostasis. Self-renewal epidermoid epithelia are continuously exposed to mutagens and are the most common target of cancer. Unknown mechanisms commit rapidly proliferating cells to post-mitotic terminal differentiation. We have over-activated or inhibited the endogenous DNA damage response (DDR) pathways by combinations of activating TopBP1 protein, specific shRNAs, or chemical inhibitors for ATR, ATM, and/or DNA-PK. The results dissect and demonstrate that these signals control keratinocyte differentiation in proliferating cells independently of actual DNA damage. The DDR limits keratinocyte multiplication upon hyperproliferative stimuli. Moreover, knocking down H2AX, a common target of the DDR pathways, inhibits the epidermoid phenotype. The results altogether show that the DDR is required to maintain the balance proliferation differentiation and suggest that is part of the squamous program. We propose a homeostatic model where genetic damage is automatically and continuously cleansed by cell-autonomous mechanisms.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Paraformaldehyde, powder, 95%
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
5-Bromo-2′-deoxyuridine, ≥99% (HPLC)
Sigma-Aldrich
Rhodamine B, ≥95% (HPLC)
Sigma-Aldrich
MISSION® pLKO.1-puro Empty Vector Control Plasmid DNA, Contains no shRNA insert
Sigma-Aldrich
Saponin Quillaja sp., Sapogenin content 20-35 %
Sigma-Aldrich
Monoclonal Anti-γ-Tubulin antibody produced in mouse, clone GTU-88, ascites fluid
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, from mouse
Sigma-Aldrich
Anti-BrdU antibody, Mouse monoclonal, clone BU-33, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Involucrin antibody produced in mouse, clone SY5, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Cytokeratin Peptide 8 antibody produced in mouse, clone M20, ascites fluid