  • Search Results
  • Antiprotozoal polyacetylenes from the Tanzanian medicinal plant Cussonia zimmermannii.
Antiprotozoal polyacetylenes from the Tanzanian medicinal plant Cussonia zimmermannii.
Journal of natural products (2007-10-10)
Martin Senn, Sigmund Gunzenhauser, Reto Brun, Urs Séquin
ABSTRACT
From the petroleum ether extract of the root bark of Cussonia zimmermannii four polyacetylenes, 1- 4, were isolated, three of which ( 1- 3) were active against Trypanosoma brucei rhodesiense, Trypanosoma cruzi, Plasmodium falciparum, and Leishmania donovani.
Sigma-Aldrich
Miltefosine, ≥98% (perchloric acid titration)
Sigma-Aldrich
Podophyllotoxin