Merck
Gallic acid-based indanone derivatives as anticancer agents.
Bioorganic & medicinal chemistry letters (2008-07-01)
Hari Om Saxena, Uzma Faridi, Suchita Srivastava, J K Kumar, M P Darokar, Suaib Luqman, C S Chanotiya, Vinay Krishna, Arvind S Negi, S P S Khanuja
ABSTRACT
Gallic acid-based indanone derivatives have been synthesised. Some of the indanones showed very good anticancer activity in MTT assay. Compounds 10, 11, 12 and 14 possessed potent anticancer activity against various human cancer cell lines. The most potent indanone (10, IC(50)=2.2 microM), against MCF-7, that is, hormone-dependent breast cancer cell line, showed no toxicity to human erythrocytes even at higher concentrations (100 microg/ml, 258 microM). While, indanones 11, 12 and 14 showed toxicities to erythrocytes at higher concentrations.
Sigma-Aldrich
Podophyllotoxin