  • Search Results
  • Antifeedant activity of spin-labeled derivatives of deoxypodophyllotoxin against Brontispa longissima larvae.
Antifeedant activity of spin-labeled derivatives of deoxypodophyllotoxin against Brontispa longissima larvae.
Natural product communications (2013-03-22)
Gang Feng, Jing Zhang, Liu Yang, Ying-Qian Liu, Zhi-Wei Zhang, Xuan Tian, Qi-An Jin, Zheng-Qiang Peng
ABSTRACT
To continue our search for natural product-based compounds for the control of Brontispa longissima larvae, eight spin-labeled podophyllotoxin derivatives (4a-h) and the intermediates 2 and 3 were tested for their insect antifeedant activity against third-instar larvae of B. longissima. Among all the tested compounds, 4a, 4c, 2 and 3 showed pronounced antifeedant activities with AFC50 values of 0.16, 0.31, 0.15 and 0.28 mg/mL, respectively. The different antifeedant activity ranges of these compounds indicated that variation of the structures of L-amino acids in these compounds markedly affected the activity profiles of this compound class, and some important SAR information has been revealed from it.
Sigma-Aldrich
Picropodophyllotoxin, ≥96% (HPLC), powder
Sigma-Aldrich
Podophyllotoxin