• Strona główna
  • Wyniki wyszukiwania
  • In utero exposure to perfluorooctanoate (PFOA) or perfluorooctane sulfonate (PFOS) did not increase body weight or intestinal tumorigenesis in multiple intestinal neoplasia (Min/+) mice.
In utero exposure to perfluorooctanoate (PFOA) or perfluorooctane sulfonate (PFOS) did not increase body weight or intestinal tumorigenesis in multiple intestinal neoplasia (Min/+) mice.
Environmental research (2014-05-20)
Ha Thi Ngo, Ragna Bogen Hetland, Azemira Sabaredzovic, Line Småstuen Haug, Inger-Lise Steffensen
ABSTRAKT
We examined whether perfluorooctanoate (PFOA) or perfluorooctane sulfonate (PFOS) had obesogenic effects and if they increased spontaneous intestinal tumorigenesis in the mouse model C57BL/6J-Min/+ (multiple intestinal neoplasia) after in utero exposure. The dams were exposed to PFOA or PFOS (0.01, 0.1 or 3.0mg/kg bw/day) by po gavage on GD1-17. The Min/+ and wild-type offspring were terminated at week 11 for examination of intestinal tumorigenesis or at week 20 for obesogenic effect, respectively. Body weights of the dams and pups were recorded throughout life. Food intake was determined at week 6 and 10. Blood glucose (non-fasted) was measured at week 6 and 11. No obesogenic effect of PFOA or PFOS was observed up to 20 weeks of age. PFOA or PFOS did not increase the incidence or number of tumors in the small intestine or colon of the Min/+ mice or affect their location along the intestines. Feed intake was not affected. There were some indications of toxicity of PFOA, but not of PFOS. There was lower survival of pups after 3.0mg/kg PFOA, lower body weight in pups after 3.0 and possibly 0.1mg/kg PFOA, and increased relative liver weight after 0.01 and possibly 0.1mg/kg PFOA. Plasma glucose was lower after 0.01 and 0.1mg/kg PFOA. In conclusion, exposure to PFOA and PFOS in utero with the doses used did not have obesogenic effect on either Min/+ or wild-type mice, at least not up to 11 or 20 weeks of age, nor increased intestinal tumorigenesis in Min/+ mice.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Perfluorooctanoic acid, 95%
Sigma-Aldrich
Heptadecafluorooctanesulfonic acid solution, ~40% in H2O (T)
Supelco
Heptadecafluorooctanesulfonic acid solution, 100 μg/mL in methanol, analytical standard
Supelco
Pentadecafluorooctanoic acid, analytical standard
Supelco
Pentadecafluorooctanoic acid solution, 100 μg/mL in methanol, analytical standard
Sigma-Aldrich
Pentadecafluorooctanoic acid ammonium salt, ≥98.0% (NT)