Merck
Rictor is required for early B cell development in bone marrow.
PloS one (2014-08-02)
Yingchi Zhang, Tianyuan Hu, Chunlan Hua, Jie Gu, Liyan Zhang, Sha Hao, Haoyue Liang, Xiaomin Wang, Weili Wang, Jing Xu, Hanzhi Liu, Bin Liu, Tao Cheng, Weiping Yuan
ABSTRACT
The development of early B cells, which are generated from hematopoietic stem cells (HSCs) in a series of well-characterized stages in bone marrow (BM), represents a paradigm for terminal differentiation processes. Akt is primarily regulated by phosphorylation at Thr308 by PDK1 and at Ser473 by mTORC2, and Akt signaling plays a key role in hematopoiesis. However, the role of mTORC2 in the development of early B cells remains poorly understood. In this study, we investigated the functional role of mTORC2 by specifically deleting an integral component, Rictor, in a hematopoietic system. We demonstrated that the deletion of Rictor induced an aberrant increase in the FoxO1 and Rag-1 proteins in BM B cells and that this increase was accompanied by a significant decrease in the abundance of B cells in the peripheral blood (PB) and the spleen, suggesting impaired development of early B cells in adult mouse BM. A BM transplantation assay revealed that the B cell differentiation defect induced by Rictor deletion was not affected by the BM microenvironment, thus indicating a cell-intrinsic mechanism. Furthermore, the knockdown of FoxO1 in Rictor-deleted HSCs and hematopoietic progenitor cells (HPCs) promoted the maturation of B cells in the BM of recipient mice. In addition, we revealed that treatment with rapamycin (an mTORC1 inhibitor) aggravated the deficiency in B cell development in the PB and BM. Taken together, our results provide further evidence that Rictor regulates the development of early B cells in a cell-intrinsic manner by modifying the expression of FoxO1 and Rag-1.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-Human IgM (μ-chain specific) antibody produced in mouse, clone MB-11, ascites fluid
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Supelco
Rapamycin, VETRANAL®, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Monoclonal Anti-Human IgM (μ-chain specific)−Alkaline Phosphatase antibody produced in mouse, clone MB-11, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus