  • Search Results
  • Haematocrit corrected analysis of creatinine in dried blood spots through potassium measurement.
Haematocrit corrected analysis of creatinine in dried blood spots through potassium measurement.
Analytical and bioanalytical chemistry (2014-11-07)
Jeroen C G den Burger, Aabraham J Wilhelm, Abdel C Chahbouni, Rene M Vos, Arno Sinjewel, Eleonora L Swart
ABSTRACT
We developed a method for the analysis of creatinine in dried blood spot (DBS) samples to facilitate monitoring of renal function in combination with TDM of immunosuppressive drugs for transplant patients outside the hospital. An 8-mm disc of the DBS was punched, extracted and followed by LC-MS/MS analysis. The haematocrit proved to have a significant influence on the analysis of creatinine in DBS samples. As potassium is a suitable marker for haematocrit, we implemented a method for measuring potassium in DBS and correct the creatinine for haematocrit. For both creatinine and K(+) in DBS analytical and DBS, validation was performed, both components met the validation criteria and no other influences beside the haematocrit were detected. To assess the haematocrit correction, samples were compared to the 'golden' standard and plotted before and after correction. The correction showed a great improvement in agreement between the DBS assay and venous blood assay.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Potassium, ingot, 99.95% trace metals basis
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Methanol solution, suitable for HPLC, contains 10 % (v/v) water, 0.1 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Potassium, cubes (in mineral oil), 99.5% trace metals basis
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard