Merck
Phytic acid: an alternative root canal chelating agent.
Journal of endodontics (2014-12-03)
Mohannad Nassar, Noriko Hiraishi, Yukihiko Tamura, Masayuki Otsuki, Kazuhiro Aoki, Junji Tagami
ABSTRACT
The objectives of this study were to investigate the effect of phytic acid, inositol hexakisphosphate (IP6), as a final rinse on the surface of instrumented root canals and smear-layered flat dentin surfaces treated with sodium hypochlorite (NaOCl) and to evaluate its effect on the viability and alkaline phosphatase activity of osteoblast-like cells (MC3T3-E1). The universally accepted chelating agent EDTA was used as the control in all conducted experiments. Root canals of human canines were instrumented with rotary files and irrigated with 5% NaOCl, followed by a final rinse of 17% EDTA (1 minute), 1% IP6 (1 minute or 30 seconds), or distilled water. NaOCl-treated flat coronal dentin surfaces were also treated with 17% EDTA (1 minute), 1% IP6 (1 minute or 30 seconds), or distilled water. The presence or absence of smear layer was evaluated with scanning electron microscopy. Cell viability and alkaline phosphatase assays were performed to evaluate the effect of IP6 and EDTA on cultured MC3T3-E1 cells. The results demonstrated the ability of IP6 to remove the smear layer from instrumented root canals and flat coronal dentin surfaces. When compared with EDTA, IP6 was less cytotoxic and did not affect the differentiation of MC3T3-E1 cells. IP6 shows the potential to be an effective and biocompatible chelating agent.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Supelco
Hexamethyldisilazane, for GC derivatization, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Hexamethyldisilazane, produced by Wacker Chemie AG, Burghausen, Germany, ≥97.0% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Hexamethyldisilazane, ReagentPlus®, 99.9%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Hexamethyldisilazane, reagent grade, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)