  • Search Results
  • Asymmetric dimethylarginine in somatically healthy schizophrenia patients treated with atypical antipsychotics: a case-control study.
Asymmetric dimethylarginine in somatically healthy schizophrenia patients treated with atypical antipsychotics: a case-control study.
BMC psychiatry (2015-04-17)
Anders Jorgensen, Ulla Knorr, Mia Greisen Soendergaard, Jens Lykkesfeldt, Anders Fink-Jensen, Henrik Enghusen Poulsen, Martin Balslev Jorgensen, Niels Vidiendal Olsen, Jonatan Myrup Staalsø
ABSTRACT
Schizophrenia is associated with increased cardiovascular morbidity and mortality. Asymmetric dimethylarginine (ADMA), an endogenous inhibitor of the nitric oxide synthase, and the L-arginine:ADMA ratio are markers of endothelial dysfunction that predict mortality and adverse outcome in a range of cardiovascular disorders. Increased ADMA levels may also lead to increased oxidative stress. We hypothesized that ADMA and the L-arginine:ADMA ratio are increased in somatically healthy schizophrenia patients treated with atypical antipsychotics (AAP), and that the ADMA and the L-arginine: ADMA ratio are positively correlated to measures of oxidative stress. We included 40 schizophrenia patients treated with AAP, but without somatic disease or drug abuse, and 40 healthy controls. Plasma concentrations of ADMA and L-arginine were determined by high-performance liquid chromatography. Data were related to markers of systemic oxidative stress on DNA, RNA and lipids, as well as measures of medication load, duration of disease and current symptomatology. Plasma ADMA and the L-arginine:ADMA ratio did not differ between schizophrenia patients and controls. Furthermore, ADMA and the L-arginine:ADMA ratio showed no correlations with oxidative stress markers, medication load, or Positive and Negative Syndrome Scale scores. Schizophrenia and treatment with AAP was not associated with increased levels of plasma ADMA or the L-arginine:ADMA ratio. Furthermore, plasma levels of ADMA were not associated with levels of systemic oxidative stress in vivo.
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Potassium, ingot, 99.95% trace metals basis
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Potassium, cubes (in mineral oil), 99.5% trace metals basis
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material