Distribution of perfluorinated compounds in Lake Taihu (China): impact to human health and water standards.
The Science of the total environment (2013-12-12)
Gang Pan, Qin Zhou, Xuan Luan, Q Shiang Fu
ABSTRAKT
The distribution in water and sediment, the sources/sinks and the risk of perfluorinated compounds (PFCs) in Lake Taihu, China were investigated. The total PFCs concentration was 164 to 299 ng L(-1) in water and 5.8 to 35 ng g(-1) (dw) in sediment. The highest concentrations of perfluorooctane sulfonate (PFOS) and perfluorooctanoate (PFOA) in water were 29.2 ng L(-1) and 136ngL(-1). PFOS was largely associated with sediment, whereas short chain PFCs predominated in water. The partition coefficient (Kd) was positively correlated with the organic carbon fraction (ƒoc) for PFOS but not for the other PFCs. The organic carbon normalized partition coefficient (Koc) increased by 0.51 log units for each additional CF2 moiety from perfluoro-butanesulfonate (PFBS) to PFOS. For the same chain length but different functional groups, the log Koc of PFOS was 1.35 units higher than PFOA. PFOS exhibited the highest affinity for sediment through the partition mechanism, and ƒoc affected the sediment as a sink of PFOS. Although there was no immediate health impact by the intake of the water alone, the consumption of aquatic products may cause potential health risks for animals/humans on the time scale of months to years. The relationship between the concentration, water-sediment distribution, bioaccumulation and toxicity should be considered in determining the water standards of PFCs.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Heptadecafluorooctanesulfonic acid solution, ~40% in H2O (T)
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Heptadecafluorooctanesulfonic acid solution, 100 μg/mL in methanol, analytical standard
Sigma-Aldrich
Potassium, cubes (in mineral oil), 99.5% trace metals basis
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, suitable for HPLC, contains 10 % (v/v) water, 0.1 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C