  • Search Results
  • Purple grape juice supplementation in smokers and antioxidant status according to different types of GST polymorphisms.
Purple grape juice supplementation in smokers and antioxidant status according to different types of GST polymorphisms.
Journal of clinical biochemistry and nutrition (2015-02-14)
Mi-Ran Cho, Jeong-Hwa Han, Hye-Jin Lee, Yoo Kyoung Park, Myung-Hee Kang
ABSTRACT
DNA damages and antioxidant status was assessed after 8 weeks of purple grape juice supplementation in male smokers depending on the glutathione S-transferase polymorphisms. Ninety-five smokers consumed 480 ml of purple grape juice for 8 weeks. The blood samples were collected before and after supplementation to measure lymphocyte DNA damages, plasma antioxidants, conjugated diene, and the erythrocyte antioxidant enzymes. The diastolic pressure, lymphocyte DNA damage, and plasma conjugated diene were significantly decreased but the plasma γ-tocopherol was increased in GSTM1-null genotype, while increased blood glutathione and decreased lymphocyte DNA damage were observed in GSTM1-present genotype. In case of GSTT1 on the other hand, the decrease in diastolic pressure and lymphocyte DNA damage was observed in both null types and present types, but the erythrocyte catalase activity was decreased in GSTT1-null type and the plasma vitamin C level was increased in GSTT1-present type, suggesting that, the antioxidant effect of grape juice was greater in GSTT1-present type compared to GSTT1-null type. The intakes of 8-week purple grape juice affected diastolic blood pressures, DNA damage reductions and antioxidant status in smokers, mainly greater in GSTM1-null type and GSTT1-present type.
Product Description
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Glutathione, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
2-Nitrobenzoic acid, 95%, Contains 3- and 4-isomers
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethidium bromide, BioReagent, for molecular biology, powder
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications