  • Search Results
  • Molecular dynamic simulation and spectroscopic investigation of some cytotoxic palladium(II) complexes interaction with human serum albumin.
Molecular dynamic simulation and spectroscopic investigation of some cytotoxic palladium(II) complexes interaction with human serum albumin.
Combinatorial chemistry & high throughput screening (2014-09-30)
Mahboube Eslami Moghadam, Maryam Saidifar, Faramarz Rostami-Charati, Davoud Ajloo, Maryam Ghadamgahi
ABSTRACT
Studies on the interactions between metallodrugs and human serum albumin (HSA), as carrier for drugs and biological molecules, are extremely important to design and discover new drugs. The interaction of three novel synthesized complexes of [Pd(phen)(R-gly)]NO3, where R-gly is methyl-, propyl-, and amyl-glycine and phen is 1,10- phenanthroline, with HSA were investigated using spectroscopic studies in combination with a molecular dynamic simulation. These water soluble complexes can denature HSA at ~50 µM. According to the results obtained for the isothermal titration at 27 and 37°C, it was found that there are 10, 8, and 6 binding sites (g) for methyl-, propyl-, and amyl-glycine complexes on the HSA with positive cooperativity in binding, respectively. Also, the binding and thermodynamic parameters were analyzed. We found a good consistency between secondary structure and simulation data with spectroscopic studies, and the experimental data are confirmed by molecular simulation results. In addition, the results related to helix, beta sheets, and coil percentages revealed that all complexes decrease the helix structure and increase the beta structure; and that the amyl derivative is more effective in denaturing the HSA structure.
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, beads, 20-40 mesh, reagent grade, 97%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%