  • Search Results
  • Improved pharmacokinetic and biodistribution properties of the selective urokinase inhibitor PAI-2 (SerpinB2) by site-specific PEGylation: implications for drug delivery.
Improved pharmacokinetic and biodistribution properties of the selective urokinase inhibitor PAI-2 (SerpinB2) by site-specific PEGylation: implications for drug delivery.
Pharmaceutical research (2014-09-19)
Kara Lea Vine, Sergei Lobov, Vineesh Indira Chandran, Nathanial Lachlan Ewart Harris, Marie Ranson
ABSTRACT
Overexpression of the serine protease urokinase (uPA) is recognised as an important biomarker of metastatic disease and a druggable anticancer target. Plasminogen activator inhibitor type-2 (PAI-2/SerpinB2) is a specific uPA inhibitor with proven potential for use in targeted therapy. However, PAI-2 is rapidly cleared via the renal system which impairs tumor uptake and efficacy. Here we aimed to improve the pharmacological properties of PAI-2 by site-specific PEGylation. Several cysteine to serine substitution mutants were generated for PEGylation with PEG-maleimide (size range 12-30 kDa) and the physico-chemical and biochemical properties of the PEG-PAI-2 conjugates characterised. Radiolabeled proteins were used for evaluation of blood clearance and tissue uptake profiles in an orthotopic breast tumor xenograft mouse model. PEGylation of the PAI-2(C161S) mutant gave a predominant mono-PEGylated-PAI-2 product (~90%) with full uPA inhibitory activity, despite a significant increase in hydrodynamic radius. Compared to un-PEGylated protein the plasma half-life and AUC for PEG20-PAI-2(C161S) were significantly increased. This translated to a 10-fold increase in tumor retention after 24 h compared to PAI-2(C161S), an effect not seen in non-target organs. Our data underscores the potential for PEG20-PAI-2(C161S) drug conjugates to be further developed as anti-uPA targeted therapeutics with enhanced tumor retention.
Sigma-Aldrich
Trypan Blue, Dye content 60 %
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Maleimide, 99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Trypan Blue solution, 0.4%, for microscopy
Ampicillin, anhydrous, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Sigma-Aldrich
Ultrapure Acetonitrile
SAFC
Isopropyl β-D-1-thiogalactopyranoside