Construction of a pIX-modified Adenovirus Vector Able to Effectively Bind to Nanoantibodies for Targeting.
Acta naturae (2014-08-06)
M N Garas, S V Tillib, O V Zubkova, V N Rogozhin, T I Ivanova, L A Vasilev, D Yu Logunov, M M Shmarov, I L Tutykhina, I B Esmagambetov, I Yu Gribova, A S Bandelyuk, B S Naroditsky, A L Gintsburg
ABSTRAKT
Current targeting strategies for genetic vectors imply the creation of a specific vector for every targeted receptor, which is time-consuming and expensive. Therefore, the development of a universal vector system whose surface can specifically bind molecules to provide efficient targeting is of particular interest. In this study, we propose a new approach in creating targeted vectors based on the genome of human adenovirus serotype 5 carrying the modified gene of the capsid protein pIX (Ad5-EGFP-pIX-ER): recombinant pseudoadenoviral nanoparticles (RPANs). The surfaces of such RPANs are able to bind properly modified chimeric nanoantibodies that specifically recognize a particular target antigen (carcinoembryonic antigen (CEA)) with high affinity. The efficient binding of nanoantibodies (aCEA-RE) to the RPAN capsid surfaces has been demonstrated by ELISA. The ability of the constructed vector to deliver target genes has been confirmed by experiments with the tumor cell lines A549 and Lim1215 expressing CEA. It has been shown that Ad5-EGFP-pIX-ER carrying aCEA-RE on its surface penetrates into the tumor cell lines A549 and Lim1215 via the CAR-independent pathway three times more efficiently than unmodified RPAN and Ad5-EGFP-pIX-ER without nanoantibodies on the capsid surface. Thus, RPAN Ad5-EGFP-pIX-ER is a universal platform that may be useful for targeted gene delivery in specific cells due to "nanoantibody-modified RPAN" binding.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-HA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-HA antibody produced in mouse, clone HA-7, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-HA antibody produced in mouse, clone HA-7, ascites fluid
Sigma-Aldrich
ANTI-CEA (CENTER) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-CTSE antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
ANTI-RCV1 antibody produced in mouse, clone 5A6, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
ANTI-RCV1 antibody produced in mouse, clone 2H4, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
ANTI-RCV1 antibody produced in mouse, clone 4H2, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
ANTI-RCV1 antibody produced in mouse, clone 4C10, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
ANTI-RCV1 antibody produced in mouse, clone 4D9, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
ANTI-RCV1 antibody produced in mouse, clone 2B3, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
ANTI-RCV1 antibody produced in mouse, clone 1D3, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
ANTI-RCV1 antibody produced in mouse, clone 4E2, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
ANTI-RCV1 antibody produced in mouse, clone 4A8, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
ANTI-RCV1 antibody produced in mouse, clone 2B2, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
ANTI-RCV1 antibody produced in mouse, clone 1B9, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
ANTI-RCV1 antibody produced in mouse, clone 1B7, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-HA antibody produced in mouse, clone 1B8, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-HA antibody produced in mouse, clone 4D4, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-HA antibody produced in mouse, clone 1F7, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-HA antibody produced in mouse, clone 3C6, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-HA antibody produced in mouse, clone 3F6, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-RCVRN antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-CTSE antibody produced in mouse, clone 2D5, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-CTSE antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.