  • Search Results
  • Urinary neutrophil gelatinase-associated lipocalin and clinical outcomes in chronic kidney disease patients.
Urinary neutrophil gelatinase-associated lipocalin and clinical outcomes in chronic kidney disease patients.
Clinical chemistry and laboratory medicine (2014-08-26)
Hugo You-Hsien Lin, Daw-Yang Hwang, Su Chu Lee, Hung-Tien Kuo, Mei-Chuan Kuo, Jer-Ming Chang, Jer-Chia Tsai, Chi-Chih Hung, Shang-Jyh Hwang, Hung-Chen Chen
ABSTRACT
Tubulointerstitial damage is a final common pathway of most renal diseases. Whether urinary neutrophil gelatinase-associated lipocalin (uNGAL), a biomarker for renal tubular damage, is of prognostic value for clinical outcomes in chronic kidney disease (CKD) patients has not been well investigated. The uNGAL and proteinuria levels were measured among a cohort of 473 advanced CKD patients of various etiologies recruited during 2002-2009. The estimated glomerular filtration rate (eGFR) was 32.3±22.0 mL/min/1.73 m2 with a urine protein-to-creatinine ratio (UPCR) 680 (255-1248) mg/g and 132 (27.9%) participants had diabetes. The baseline uNGAL level was significantly associated with male gender, eGFR, UPCR, and hemoglobin. The hazard ratio (HR) of the highest uNGAL tertile for end-stage renal disease (ESRD) was 3.44 (95% CI 1.47-8.06, p=0.004). With the adjustment of urine creatinine and urine protein, HR of the highest urine NGAL-to-creatinine ratio (UNCR) tertile and the highest urine NGAL-to-protein ratio (UNPR) tertile was 3.06 (95% CI 1.19-7.90, p=0.02) and 2.10 (95% CI 1.13-3.89, p=0.02), respectively. UNPR increased the prediction of survival model for ESRD. HR of the highest UNCR tertile and UNPR tertile for cardiovascular (CV) events was 2.21 (95% CI 0.81-5.98, p=0.08) and 2.79 (95% CI 1.25-6.26, p=0.01), respectively. None of these were associated with all-cause mortality. Elevated uNGAL in CKD patients is associated with risks for ESRD and probably CV events. UNPR could improve the prediction for ESRD.
Product Description
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Potassium, ingot, 99.95% trace metals basis
Sigma-Aldrich
Uric acid, ≥99%, crystalline
Sigma-Aldrich
Uric acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Potassium, cubes (in mineral oil), 99.5% trace metals basis
SAFC
Cholesterol, Plant-Derived, SyntheChol®, NF, Ph. Eur., JP
Sigma-Aldrich
Potassium hydride, in paraffin