  • Search Results
  • Abnormal sinoatrial node development resulting from disturbed vascular endothelial growth factor signaling.
Abnormal sinoatrial node development resulting from disturbed vascular endothelial growth factor signaling.
International journal of cardiology (2015-02-24)
Emmeline E Calkoen, Rebecca Vicente-Steijn, Nathan D Hahurij, Conny J van Munsteren, Arno A W Roest, Marco C DeRuiter, Paul Steendijk, Martin J Schalij, Adriana C Gittenberger-de Groot, Nico A Blom, Monique R M Jongbloed
ABSTRACT
Sinus node dysfunction is frequently observed in patients with congenital heart disease (CHD). Variants in the Vascular Endothelial Growth Factor-A (VEGF) pathway are associated with CHD. In Vegf(120/120) mice, over-expressing VEGF120, a reduced sinoatrial node (SAN) volume was suggested. Aim of the study is to assess the effect of VEGF over-expression on SAN development and function. Heart rate was measured in Vegf(120/120) and wildtype (WT) embryos during high frequency ultrasound studies at embryonic day (E)12.5, 14.5 and 17.5 and by optical mapping at E12.5. Morphology was studied with several antibodies. SAN volume estimations were performed, and qualitative-PCR was used to quantify expression of genes in SAN tissues of WT and Vegf(120/120) embryos. Heart rate was reduced in Vegf(120/120) compared with WT embryos during embryonic echocardiography (52 ± 17 versus 125 ± 31 beats per minute (bpm) at E12.5, p<0.001; 123 ± 37 vs 160 ± 29 bmp at E14.5, p=0.024; and 177 ± 30 vs 217 ± 34 bmp, at E17.5 p=0.017) and optical mapping (81 ± 5 vs 116 ± 8 bpm at E12.5; p=0.003). The SAN of mutant embryos was smaller and more vascularized, and showed increased expression of the fast conducting gap junction protein, Connexin43. Over-expression of VEGF120 results in reduced heart rate and a smaller, less compact and hypervascularized SAN with increased expression of Connexin43. This indicates that VEGF is necessary for normal SAN development and function.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Potassium, ingot, 99.95% trace metals basis
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Supelco
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Potassium, cubes (in mineral oil), 99.5% trace metals basis