  • Search Results
  • Physiological responses and characteristics of sperm collected after electroejaculation or transrectal ultrasound-guided massage of the accessory sex glands in anesthetized mouflons (Ovis musimon) and Iberian ibexes (Capra pyrenaica).
Physiological responses and characteristics of sperm collected after electroejaculation or transrectal ultrasound-guided massage of the accessory sex glands in anesthetized mouflons (Ovis musimon) and Iberian ibexes (Capra pyrenaica).
Theriogenology (2015-07-16)
Rodolfo Ungerfeld, Antonio López-Sebastián, Milagros Esteso, Jorgea Pradiee, Adolfo Toledano-Díaz, Cristina Castaño, Beatriz Labrador, Julián Santiago-Moreno
ABSTRACT
The objective was to characterize the stress response and the seminal parameters obtained with electroejaculation (EE) or transrectal ultrasound-guided massage of the accessory sex glands (TUMASG) in two captive but nondomestic ruminants, the mouflons and the Iberian ibex under general anesthesia. In mouflons, the physiological responses (heart and respiratory rate, rectal temperature, cortisol, creatine kinase, potassium and glucose concentrations) changed similarly with both procedures. The TUMASG procedure was faster than EE in mouflons (21.7 ± 1.4 vs. 12.4 ± 1.2 minutes, P < 0.01). In ibexes, respiratory rate, cortisol and creatine kinase concentration changes were greater with EE than with TUMASG (final respiratory rate: 62.7 ± 5.5 vs. 38.1 ± 5.6 breaths/min [P < 0.05]; final cortisol: 51.4 ± 5.1 vs. 25.3 ± 5.6 ng/mL [P < 0.001]; and final creatine kinase: 300.9 ± 99.9 vs. 87.1 ± 16.9 U/L [P < 0.001]). Electroejaculation provided better results in some sperm parameters (mouflons: sperm score: 3.4 ± 0.3 vs. 2.6 ± 0.2 [P < 0.01]; total number of sperm ejaculated: 982.4 ± 299 vs. 710.0 ± 542.2 [P < 0.05]; ibexes: sperm with progressive motility: 47.7 ± 6.2 vs. 20.5 ± 8.3 [P < 0.05]). The transrectal ultrasound-guided massage of the accessory sex glands appears to be an alternative technique to collect sperm from wild ruminants, reducing the need for electrical stimuli and thus decreasing the undesired responses of EE in the more sensitive species. On the other hand, better fresh sperm may be collected with EE. However, TUMASG provides practical advantages in animal welfare, firstly in these wild species more sensible to stress management and capture myopathy.
Sigma-Aldrich
Yohimbine hydrochloride, ≥98% (HPLC), powder
Sigma-Aldrich
Potassium, cubes (in mineral oil), 99.5% trace metals basis
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Potassium, ingot, 99.95% trace metals basis
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil