Current exposure of Italian women of reproductive age to PFOS and PFOA: A human biomonitoring study.
Chemosphere (2015-05-13)
Elena De Felip, Annalisa Abballe, Fulvia Lucia Albano, Tatiana Battista, Valter Carraro, Michele Conversano, Silva Franchini, Laura Giambanco, Nicola Iacovella, Anna Maria Ingelido, Antonio Maiorana, Francesco Maneschi, Valentina Marra, Antonella Mercurio, Roberta Nale, Bianca Nucci, Vincenzo Panella, Flavia Pirola, Maria Grazia Porpora, Enrico Procopio, Nicola Suma, Silvia Valentini, Luisa Valsenti, Valerio Vecchiè
ABSTRAKT
Perfluorooctane sulfonate (PFOS) and perfluorooctanoic acid (PFOA) concentrations were determined in serum samples collected in 2011-2012 from 549 nulliparous Italian women of reproductive age who resided in six different Italian Regions. Assessment of exposure to perfluorinated compounds was part of a large human biomonitoring study (Project Life Plus "Womenbiopop") that aimed at examining the exposure of women of reproductive age to priority organic pollutants. The median concentrations of PFOS and PFOA were 2.43, and 1.55 ng g(-1), respectively. Significant differences in the concentrations of both compounds were observed among the six Regions. Women from central Italy had the highest levels of both compounds, followed by women from northern Italy, and southern Italy. No differences in the PFOS concentrations were found between women from urban/industrial areas and women from rural areas, whereas the levels of PFOA were significantly higher in women residing in urban/industrial areas than in women residing in rural areas. Taken together, the observed concentrations confirm that the overall exposure of the Italian population is among the lowest observed in industrialized countries. A downward temporal trend in exposure was observed for both compounds when comparing the results from the present study with those assessed in a study conducted in 2008.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Perfluorooctanoic acid, 95%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Heptadecafluorooctanesulfonic acid solution, ~40% in H2O (T)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Sigma-Aldrich
Ultrapure Acetonitrile