Koszyk0
[Home](https://www.sigmaaldrich.com/PL/en)[Terpenes in Cannabis: Profile and Importance](https://www.sigmaaldrich.com/PL/en/applications/environmental-testing-and-industrial-hygiene/cannabis-testing/terpenes-cannabis)Headspace SPME-GC/MS Analysis of Terpenes in Hops and Cannabis
# Headspace SPME-GC/MS Analysis of Terpenes in Hops and Cannabis
__Katherine K. Stenerson, Principal Scientist__
[*Article from Analytix Reporter - Issue 3*](https://www.sigmaaldrich.com/PL/en/collections/analytix-reporter)
__Section Overview__
- [Using Terpene Profile for Plant Identification](https://www.sigmaaldrich.com#using)
- [Experimental](https://www.sigmaaldrich.com#experimental)
- [Analysis and Quantification of Terpenes by SPME GC-MS](https://www.sigmaaldrich.com#analysis)
- [SPME Method Optimization](https://www.sigmaaldrich.com#spme)
- [Identification of Terpenes Using GC/MS](https://www.sigmaaldrich.com#identitification)
- [Terpenes in Hops Samples](https://www.sigmaaldrich.com#hops)
- [Terpenes in Cannabis Sample](https://www.sigmaaldrich.com#cannibis)
- [Conclusion](https://www.sigmaaldrich.com#conclusion)
*In this application, headspace-SPME combined with GC/MS was used to analyze some of the terpenes present in both common hops and cannabis.*
Terpenes are small molecules synthesized by some plants. The name terpene is derived from turpentine, which contains high concentrations of these compounds. Terpene molecules are constructed from the joining of isoprene units in a head-to-tail configuration (__Figure 1__). Classification is then done according to the number of these isoprene units in the structure (__Table 1__). The configurations of terpenes can be cyclic or open, and can include double bonds, and hydroxyl, carbonyl or other functional groups. If the terpene contains elements other than C and H, it is referred to as a terpenoid.1
__Figure 1.__ Isoprene Unit
| | |
|----------------|--------------------------|
| Classification | Number of Isoprene Units |
| Monoterpene | 2 |
| Sesquiterpene | 3 |
| Diterpene | 4 |
| Triterpene | 6 |
| Tetraterpene | 8 |
Table 1.Classification of Terpenes
Terpenes are present in essential oils derived from plants and often impart characteristic aromas to the plant or its oil. For example, d-Limonene, which is found in lemon, orange, caraway, and other plant oils, has a lemon-like odor. Essential oils, with their component terpenes and terpenoids, have been applied in therapeutic use known as aromatherapy to aid in the relief of conditions such as anxiety, depression, and insomnia.2 This has led to the use of plants which contain these compounds in preparations such as oils, teas, and tonics.
## [](https://www.sigmaaldrich.com)Using Terpene Profile for Plant Identification
The *cannabis sativa* (cannabis or marijuana) plant contains over 100 different terpenes and terpenoids, including mono, sesqui, di, and tri, as well as other miscellaneous compounds of terpenoid orgin.3 Although the terpene profile does not necessarily indicate geographic origin of a cannabis sample, it can be used in forensic applications to determine the common source of different samples.4 In addition, different cannabis strains have been developed which have distinct aromas and flavors; a result of the differing amounts of specific terpenes present.5 *Humulus lupulus* (common hops) and cannabis are both members of the family Cannabaceae.6 Consequently, there are similarities in the terpenes each contains. Terpenes give both plant commodities characteristic organoleptic properties and, in the case of cannabis, produce characteristic aromas when the buds are heated or vaporized.7
## [](https://www.sigmaaldrich.com)Experimental
A Dried cannabis sample was obtained courtesy of Dr. Hari H. Singh, Program Director at the Chemistry & Physiological Systems Research Branch of the United States National Institute on Drug Abuse at the National Institute of Health. The extract strain of the sample was not known. Hop flowers of an unknown variety were purchased from an on-line source. Pellets of Cascade and US Golding hop varieties were purchased at a local home-brew supply shop. Chromatographic separation was performed on an Equity®-1 capillary GC column, and identification was done using retention indices and spectral library matches. Final analytical conditions appear in the figures.
## [](https://www.sigmaaldrich.com)Analysis and Quantification of Terpenes by SPME GC-MS
SPME involves the use of a fiber coated with an extracting phase, which is exposed to the sample to extract the target compounds for terpene quantification. The fiber is then directly inserted into the gas chromatograph (GC) injector, where the compounds are desorbed and separated by the GC column. The separated compounds are then detected and identified by the mass spectrometer (MS). The abundance of each terpene in the sample is determined by analyzing the peak areas or heights in the mass chromatogram. The data obtained from the SPME GC-MS analysis is then processed and analyzed to quantify the terpenes present in the sample.
## [](https://www.sigmaaldrich.com)SPME Method Optimization
The SPME method for terpene analysis was developed using a sample of dried hops flowers (0.2 g in a 10 mL vial). The initial SPME parameters were based on previously published work.8 The GC/MS results of this analysis are shown in __Figure 2__. This initial set of parameters used 100 µm PDMS fiber, a 1 g sample size, and 60-minute equilibration at room temperature prior to extraction. The sample size was then scaled down to 0.2 g, and the equilibration temperature was increased to 40 °C. This increased temperature allowed the equilibration time to be decreased from 60 to 30 minutes without a loss in sensitivity (__Figures 3 and 4__). The initial extraction time used was 20 minutes, and a shorter extraction time of 10 minutes was evaluated. However, a loss in sensitivity was noted, so the extraction time was maintained at 20 minutes. The DVB/CAR/PDMS fiber was then evaluated (__Figure 5__). As expected, this fiber extracted more of the lighter compounds, which, by MS spectral match, were identified as short-chain alcohols and acids.
")
__Figure 2.__ Headspace SPME-GC/MS Analysis of Dried Hops Flowers (100 µm PDMS Fiber, 1 g Sample)
| | |
|-----------------------|---------------------------------------------------|
| Sample/matrix: | 1 g ground hop flowers |
| SPME fiber: | 100 µm PDMS (57341-U) |
| Sample equilibration: | 60 min, room temperature |
| Extraction: | 20 min, headspace, 40 °C |
| Desorption process: | 3 min, 270 °C |
| Fiber post bake: | 3 min, 270 °C |
| Column: | Equity®-1, 60 m x 0.25 mm I.D., 0.25 µm (28047-U) |
| Oven: | 60 °C (2 min), 5 °C/min to 275 °C (5 min) |
| Inj. temp.: | 270 °C |
| Detector: | MSD |
| MSD interface: | 300 °C |
| Scan range: | full scan, m/z 50-500 |
| Carrier gas: | helium, 1 mL/min constant flow |
| Liner: | 0.75 mm ID SPME |
__Figure 3.__ Headspace SPME-GC/MS Analysis of Dried Hops Flowers (100 µm PDMS Fiber, 0.2 g Sample)
| |
|-----------------------------------------|
| Conditions same as Figure 2 except: |
| sample/matrix: 0.2 g ground hop flowers |
__Figure 4.__ Headspace SPME-GC/MS Analysis of Dried Hops Flowers, Increased Sample Equilibration Temperature (100 µm PDMS Fiber, 0.2 g Sample)
| | |
|-------------------------------------|--------------------------|
| Conditions same as Figure 2 except: | |
| sample/matrix: | 0.2 g ground hop flowers |
| sample equilibration: | 30 min, 40 °C |
__Figure 5.__ Headspace SPME-GC/MS Analysis of Dried Hops Flowers, Increased Sample Equilibration Temperature (DVB/CAR/PDMS Fiber, 0.2 g Sample)
| | |
|-------------------------------------|---------------------------------|
| Conditions same as Figure 2 except: | |
| sample/matrix: | 0.2 g ground hop flowers |
| SPME fiber: | 50/30 µm DVB/CAR/PDMS (57298-U) |
| sample equilibration: | 30 min, 40 °C |
| sample/matrix: | 0.2 g ground hop flowers |
| sample equilibration: | 30 min, 40 °C |
## [](https://www.sigmaaldrich.com)Identification of Terpenes Using GC/MS
Using the DVB/CAR/PDMS fiber, samples of hops and cannabis were analyzed using the optimized SPME method. Peak identifications were assigned using MS spectral matching against reference spectra in the Wiley and NIST libraries. Confirmatory identification was done based on retention index. Retention indices were calculated for the compounds identified in each sample using an *n*-alkane standard analyzed under the same GC conditions. This data was compared with published values (__Tables 2 and 3__), and final identifications were assigned, as shown in __Figures 6 and 7__.
## [](https://www.sigmaaldrich.com)Terpenes in Hops Samples
For the dried hop flower sample (__Figure 5__), the terpene profile should have shown a predominance of β-myrcene, humulene, and caryophyllene, which are typical aroma compounds in hops and hop oil.9 While caryophyllene was identified, both β-myrcene and humulene were not present at levels high enough to be detected by a library search. This may be due to the condition of the sample or the actual variety of hops analyzed since terpene profiles are known to vary between different hop varieties.10 The variety of the hop flowers analyzed is unknown, as the identity was not indicated on the packaging. For comparison, samples of two different varieties of pelletized hops were analyzed after grinding.
These samples appeared green in color, and had a much more characteristic hops-like odor than the dried flowers. Analysis of these samples showed a characteristic terpene profile, with high levels of β-myrcene, caryophyllene, and humulene present in both (__Figure 6__). The SPME method was able to detect differences in the terpene profiles between the two hops varieties. For example, farnesene (peak 18) was identified in the Cascade hops, but was too low to be confirmed in the US Goldings sample. The level of farnesene in Cascade hops is expected to be 3-7% of total oils, while in US Goldings the level should be <1%.13
## [](https://www.sigmaaldrich.com)Terpenes in Cannabis Sample
The terpenes identified in the cannabis sample (__Figure 7__) are indicated in Table 3. The profile was similar to those found previously in the analysis of dried cannabis.4,8 Peaks 1-27 in __Figure 7__ (with the exception of peak 7) were monoterpenes and monoterpenoids. The later eluting peaks consisted of sequiterpenes and caryophyllene oxide, which is a sequiterpenoid. The most abundant terpene was caryophyllene. The predominance of this compound could be due to the specific strain of cannabis tested, and/or the nature of the sample tested, which was dried. Previous studies have shown the level of this compound to increase significantly relative to other terepenes and terpenoids with drying.4 Consequently, the levels of the more volatile monoterpenes and terpenoids would be expected to be less, and this was observed to some degree. Among the monoterpenes and terpenoids the most abundant were α-pinene and d-Limonene.
| | | | | | |
|----------|----------|----------------------------------|-----------------|-----------------|-----------|
| Peak No. | RT (min) | Name | RI (calculated) | RI (literature) | Reference |
| 1 | 8.58 | Hexanal | — | 780 | 11 |
| 2 | 12.84 | α-Pinene | 939 | 942 | 11 |
| 3 | 13.28 | Camphene | 953 | 954 | 11 |
| 4 | 13.71 | 6-Methyl-5- hepten-2-one | 966 | 968 | 11 |
| 5 | 14.1 | β-Pinene | 979 | 981 | 11 |
| 6 | 14.41 | β-Myrcene | 988 | 986 | 11 |
| 7 | 15.32 | Cymene | 1018 | 1020 | 11 |
| 8 | 15.65 | d-Limonene | 1030 | 1030 | 11 |
| 9 | 15.98 | β-Ocimene | 1041 | 1038 | 11 |
| 10 | 16.72 | *cis*-Linalool oxide | 1066 | 1068 | 11 |
| 11 | 17.49 | Linalool | 1089 | 1092 | 11 |
| 12 | 21.86 | Geraniol | 1239 | 1243 | 11 |
| 13 | 25.28 | Geranyl acetate | 1363 | 1364 | 11 |
| 14 | 25.85 | α-Ylangene | 1384 | 1373 | 8 |
| 15 | 25.97 | α-Copaene | 1388 | 1398 | 11 |
| 16 | 27.22 | Caryophyllene | 1437 | 1428 | 11 |
| 17 | 27.4 | *trans*-α- Bergamotene + unknown | 1445 | 1443 | 12 |
| 18 | 17.63 | *trans*-β-Farnesene | 1454 | 1450 | 8 |
| 19 | 28.11 | Humulene | 1473 | 1465 | 11 |
| 20 | 28.41 | γ-Muurolene | 1484 | 1475 | 11 |
| 21 | 28.45 | γ-Selinene | 1486 | 1472 | 12 |
| 22 | 28.68 | Geranyl isobutyrate | 1495 | 1493 | 11 |
| 23 | 28.79 | β-Selinene | 1499 | 1487 | 8 |
| 24 | 28.94 | α-Muurolene | 1505 | 1500 | 11 |
| 25 | 28.97 | α-Selinene | 1507 | 1501 | 12 |
| 26 | 29.31 | γ-Cadinene | 1521 | 1518 | 11 |
| 27 | 29.37 | Calamenene | 1524 | 1518 | 11 |
| 28 | 29.45 | Δ-Cadinene | 1527 | 1524 | 11 |
| 29 | 30.93 | Caryophyllene oxide | 1590 | 1584 | 8 |
| 30 | 31.5 | Humulene oxide | 1614 | 1599 | 12 |
Table 2.Terpenes in Hops Pellets Identified by MS Spectral Library Match and Retention Index.
__Figure 6.__ Headspace SPME-GC/MS Analysis of Hops Pellets Using Final Optimized Method
| | |
|--------------------------------------------------|-----------------------------------------|
| The peak elution order is listed in __Table 2.__ | |
| Conditions same as __Figure 2__ except: | |
| sample/matrix: | 0.5 g ground hop flowers (hops pellets) |
| SPME fiber: | 50/30 µm DVB/CAR/PDMS (57298-U) |
| sample equilibration: | 30 min, 40 °C |
__Figure 7.__ Headspace SPME-GC/MS Analysis of Dried Cannabis Using Final Optimized Method
| | |
|--------------------------------------------------|---------------------------------|
| The peak elution order is listed in __Table 3__. | |
| Same as __Figure 2__ except: | |
| sample/matrix: | 0.5 g dried, ground cannabis |
| SPME fiber: | 50/30 µm DVB/CAR/PDMS (57298-U) |
| sample equilibration: | 30 min, 40 °C |
| | | | | | |
|----------|----------|----------------------------------|-----------------|-----------------|-----------|
| Peak No. | RT (min) | Name | RI (calculated) | RI (literature) | Reference |
| 1 | 8.57 | Hexanal | — | — | — |
| 2 | 10.05 | Hexene-1-ol | — | — | — |
| 3 | 10.89 | 2-Heptanone | — | — | — |
| 4 | 12.56 | α-Thujene | 928 | 938 | 11 |
| 5 | 12.86 | α-Pinene + unknown | 939 | 942 | 11 |
| 6 | 13.27 | Camphene | 953 | 954 | 11 |
| 7 | 13.69 | 6-Methyl-5- hepten-2-one | 966 | 968 | 11 |
| 8 | 14.09 | β-Pinene | 979 | 981 | 11 |
| 9 | 14.27 | β-Myrcene | 984 | 986 | 11 |
| 10 | 15.09 | δ-3-Carene | 1010 | 1015 | 12 |
| 11 | 15.2 | α-Terpinene | 1014 | 1012 | 12 |
| 12 | 15.29 | Cymene | 1018 | 1020 | 11 |
| 13 | 15.6 | d-Limonene | 1028 | 1030 | 11 |
| 14 | 16.42 | γ-Terpinene | 1056 | 1057 | 11 |
| 15 | 16.6 | *trans*-Sabinene hydrate | 1062 | 1078 | 11 |
| 16 | 16.72 | *cis*-Linalool oxide | 1066 | 1068 | 11 |
| 17 | 17.43 | Linalool | 1087 | 1092 | 11 |
| 18 | 18.04 | d-Fenchyl alcohol | 1107 | 1110 | 11 |
| 19 | 18.82 | *trans*-Pinocarveol | 1135 | 1134 | 12 |
| 20 | 19.59 | Borneol L | 1161 | 1164 | 11 |
| 21 | 19.81 | 1,8-Methandien- 4-ol | 1168 | 1173 | 8 |
| 22 | 19.81 | *p*-Cymen-8-ol | 1168 | 1172 | 12 |
| 23 | 19.92 | Terpinene-4-ol | 1172 | 1185 | 11 |
| 24 | 20.22 | α-Terpineol | 1181 | 1185 | 11 |
| 25 | 24.2 | Piperitenone | 1322 | 1320 | 12 |
| 26 | 24.76 | Piperitenone oxide | 1344 | 1352 | 12 |
| 27 | 25.85 | α-Ylangene | 1384 | 1373 | 8 |
| 28 | 25.97 | α-Copaene | 1388 | 1398 | 11 |
| 29 | 26.76 | γ-Caryophyllene | 1419 | 1403 | 12 |
| 30 | 27.01 | α-Santalene | 1429 | 1428 | 12 |
| 31 | 27.16 | Caryophyllene | 1435 | 1428 | 11 |
| 32 | 27.36 | *trans*-α- Bergamotene + unknown | 1443 | 1443 | 12 |
| 33 | 27.49 | α-Guaiene | 1448 | 1441 | 8 |
| 34 | 27.56 | *trans*-β-Farnesene | 1451 | 1446 | 12 |
| 35 | 27.98 | Humulene | 1467 | 1465 | 11 |
| 36 | 28.17 | Alloaromadendrene | 1475 | 1478 | 11 |
| 37 | 28.25 | α-Curcumene | 1478 | 1479 | 12 |
| 38 | 28.75 | β-Selinene | 1497 | 1487 | 8 |
| 39 | 28.97 | α-Selinene | 1507 | 1497 | 8 |
| 40 | 28.97 | β-Bisobolene | 1507 | 1506 | 8 |
| 41 | 29.13 | α-Bulnesene | 1514 | 1513 | 12 |
| 42 | 30.12 | Selina-3,7(11)- diene | 1556 | 1542 | 12 |
| 43 | 30.94 | Caryophyllene oxide | 1590 | 1595 | 12 |
| 44 | 31.5 | Humulene oxide | 1614 | 1599 | 12 |
| 45 | 32.48 | Caryophylla-3, 8(13)-dien-5-ol A | 1658 | 1656 | 12 |
Table 3.Terpenes in Dried Cannabis Identified by MS Spectral Library Match and Retention Index
## [](https://www.sigmaaldrich.com)Conclusion
A simple headspace SPME-GC/MS method was used in the analysis of the terpene/terpenoid profiles of both hops and cannabis. The method was able to detect the characteristic terpenes and terpenoids of both, and to distinguish between different hops varieties.
[Analytix Reporter – Analytical Journal](https://www.sigmaaldrich.com/PL/en/collections/analytix-reporter)
## Related Products
Przepraszamy, wystąpił nieoczekiwany błąd
Response not successful: Received status code 500
### References
1\.
Fessenden R, Fessenden J. 1979. Lipids and Related Natural Products. Organic Chemistry. 2nd Edition. Boston, MA: Willard Grant Press.
2\.
Turner CE, Elsohly MA, Boeren EG. 1980. Constituents of Cannabis sativa L. XVII. A Review of the Natural Constituents. J. Nat. Prod.. 43(2):169-234. [https://doi.org/10.1021/np50008a001](https://doi.org/10.1021/np50008a001)
3\.
Hood L, Barry G. 1978. Headspace volatiles of marihuana and hashish: Gas chromatographic analysis of samples of different geographic origin. Journal of Chromatography A. 166(2):499-506. [https://doi.org/10.1016/s0021-9673(00)95633-4](https://doi.org/10.1016/s0021-9673%2800%2995633-4)
4\.
Rahn B. 2014. Terpenes: The Flavors of Cannabis Aromatherapy.. \[Internet].\[cited 16 Sep 2015]. Available from: [https://www.leafly.com/news](https://www.leafly.com/news)
5\.
Plants Database. \[Internet].\[cited 12 Sep 2015]. Available from: [https://plants.sc.egov.usda.gov/java/](https://plants.sc.egov.usda.gov/java/)
6\.
2015\. Cannabis Safety. Chem. Eng. News Archive. 93(16):27-28. [https://doi.org/10.1021/cen-09316-scitech1](https://doi.org/10.1021/cen-09316-scitech1)
7\.
Marchini M, Charvoz C, Dujourdy L, Baldovini N, Filippi J. 2014. Multidimensional analysis of cannabis volatile constituents: Identification of 5,5-dimethyl-1-vinylbicyclo\[2.1.1]hexane as a volatile marker of hashish, the resin of Cannabis sativa L.. Journal of Chromatography A. 1370200-215. [https://doi.org/10.1016/j.chroma.2014.10.045](https://doi.org/10.1016/j.chroma.2014.10.045)
8\.
Wolfe PH, Thesis M. 2012. Oregon State University. \[Internet].
9\.
Hop varieties. \[Internet].\[updated 13 Sep 2015]. Available from: [https://www.barthhaas.com/](https://www.barthhaas.com/)
10\.
Jennings W, Shibamoto T. 1980. Qualitative Analysis of Flavor and Fragrance Volatiles by Glass Capillary Gas Chromatography. Academic Press.
11\.
NIST Chemistry WebBook. \[Internet]. Available from: [https://webbook.nist.gov/chemistry/](https://webbook.nist.gov/chemistry/)
12\.
Variety Manual USA Hops. Hop Growers of America. \[Internet].\[cited 20 Sep 2015]. Available from: [https://www.usahops.org/](https://www.usahops.org/)
__Related Articles__
- [Analysis of Residual Solvents in Hemp Extracts](https://www.sigmaaldrich.com/PL/en/technical-documents/protocol/food-and-beverage-testing-and-manufacturing/chemical-analysis-for-food-and-beverage/residual-solvents-hemp)
- [Fast and Reliable Environmental Analysis of Aldehyde and Ketone Air Pollutants](https://www.sigmaaldrich.com/PL/en/technical-documents/protocol/analytical-chemistry/calibration-qualification-and-validation/fast-and-reliable)
- [Complete Workflow for Comprehensive Cannabis Terpenes Analysis](https://www.sigmaaldrich.com/PL/en/technical-documents/protocol/analytical-chemistry/small-molecule-hplc/cannabis-testing)
- [GC Analysis of Pyrazines in Peanut Butter](https://www.sigmaaldrich.com/PL/en/technical-documents/protocol/analytical-chemistry/solid-phase-extraction/gc-analysis-of-pyrazines-in-peanut-butter)
- [Bisphenol A Analysis in Food by SPME](https://www.sigmaaldrich.com/PL/en/technical-documents/protocol/analytical-chemistry/solid-phase-microextraction/analysis-of-bisphenol-a)
- [Developing a Quantitative SPME Method](https://www.sigmaaldrich.com/PL/en/technical-documents/protocol/analytical-chemistry/solid-phase-microextraction/developing-a-quantitative)
- [GC Analysis of Methanol Impurity in Biodiesel on Equity®-1 after SPME using 85 µm Polyacrylate Fiber](https://www.sigmaaldrich.com/PL/en/technical-documents/protocol/analytical-chemistry/solid-phase-microextraction/en-14110-gc-analysis-of-methanol-impurity-in-biodiesel-g005512)
- [GC Analysis of Aldehydes (PFBOA Derivatives) in Beer on SLB®-5ms after SPME using a 65 µm PDMS/DVB Fiber](https://www.sigmaaldrich.com/PL/en/technical-documents/protocol/analytical-chemistry/solid-phase-microextraction/gc-analysis-of-aldehydes-pfboa-derivatives-in-beer-on-slb)
- [View More](https://www.sigmaaldrich.com/PL/en/search/facet-search?focus=sitecontent&term=facet-search)
__Related Product Categories__
- [Phytochemical Standards](https://www.sigmaaldrich.com/PL/en/products/analytical-chemistry/reference-materials/phytochemical-standards)
- [Solid Phase Micro-Extraction (SPME)](https://www.sigmaaldrich.com/PL/en/products/analytical-chemistry/analytical-sample-preparation/spme-fibers-and-accessories)
Góra
__Zaloguj się, aby kontynuować__
Zaloguj się__Nie masz konta użytkownika?__Zarejestruj
Wsparcie[Obsługa klienta](https://www.sigmaaldrich.com/PL/pl/support/customer-support)[Kontakt](https://www.sigmaaldrich.com/PL/pl/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Karty charakterystyki (SDS)](https://www.sigmaaldrich.com/PL/pl/documents-search?tab=sds)[Certyfikaty (COA/COO)](https://www.sigmaaldrich.com/PL/pl/documents-search?tab=coa)[Jakość i przepisy](https://www.sigmaaldrich.com/PL/pl/life-science/quality-and-regulatory-management)[Kalkulatory i aplikacje](https://www.sigmaaldrich.com/PL/pl/support/calculators-and-apps)[Webinaria](https://www.sigmaaldrich.com/PL/pl/collections/webinars)
Zamówienia[Wyszukiwanie zamówień](https://www.sigmaaldrich.com/PL/pl/order-lookup)[Szybkie zamówienie](https://www.sigmaaldrich.com/PL/pl/quick-order)[Produkty niestandardowe](https://www.sigmaaldrich.com/PL/pl/services/custom-products)[Rozwiązania eCommerce](https://www.sigmaaldrich.com/PL/pl/life-science/ecommerce/ecommerce-solutions)
Firma[O nas](https://www.sigmaaldrich.com/PL/pl/life-science/about-us)[Odpowiedzialność](https://www.sigmaaldrich.com/PL/pl/life-science/ssbi)[Wydarzenia](https://www.sigmaaldrich.com/PL/pl/collections/events)[Komunikaty prasowe](https://www.sigmaaldrich.com/PL/pl/collections/press)[Programy](https://www.sigmaaldrich.com/PL/pl/life-science/partnership-programs)[Kariera](https://careers.merckgroup.com/global/en)[Biura](https://www.sigmaaldrich.com/PL/pl/collections/offices)
Media społecznościowe
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
Merck
__Badania. Rozwój. Produkcja.__
Jesteśmy wiodącym dostawcą dla globalnej branży Life Science, oferującym rozwiązania i usługi w zakresie badań, rozwoju i produkcji biotechnologicznej oraz rozwoju i produkcji leków farmaceutycznych.
[](https://www.sigmaaldrich.com/PL/pl)
[Rozwiązania Sigma-Aldrich®](https://www.sigmaaldrich.com/PL/pl/life-science/sigma-aldrich)[Rozwiązania BioReliance®](https://www.sigmaaldrich.com/PL/pl/life-science/bioreliance)[Roztwory Millipore®](https://www.sigmaaldrich.com/PL/pl/life-science/millipore)[Rozwiązania SAFC®](https://www.sigmaaldrich.com/PL/pl/life-science/safc)[Rozwiązania Milli-Q®](https://www.sigmaaldrich.com/PL/pl/life-science/milliq)[Rozwiązania Supelco®](https://www.sigmaaldrich.com/PL/pl/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Niemcy i/lub jej podmioty stowarzyszone. Wszelkie prawa zastrzeżone, w tym eksploracja tekstu i danych na potrzeby szkolenia w zakresie sztucznej inteligencji i podobnych technologii.
Powielanie jakichkolwiek materiałów ze strony jest surowo zabronione bez zezwolenia.
[Warunki korzystania z witryny](https://www.sigmaaldrich.com/PL/pl/life-science/legal/site-use-terms)|[Polityka prywatności](https://www.sigmaaldrich.com/PL/pl/life-science/legal/privacy-statement)|[Ogólne warunki sprzedaży](https://www.sigmaaldrich.com/PL/pl/life-science/legal/terms-and-conditions)|[Zgoda na prawa autorskie](https://www.sigmaaldrich.com/PL/pl/life-science/legal/copyright-consent)|[Mapa witryny](https://www.sigmaaldrich.com/PL/pl/site-map)