MilliporeSigma
  • Structure elucidation of a novel yellow chromophore from human lens protein.
Structure elucidation of a novel yellow chromophore from human lens protein.
The Journal of biological chemistry (2004-08-19)
Rongzhu Cheng, Qi Feng, Ognyan K Argirov, Beryl J Ortwerth
ABSTRACT
We report here the isolation of a novel acid-labile yellow chromophore from the enzymatic digest of human lens proteins and the identification of its chemical structure by liquid chromatography-mass spectrometry, liquid chromatography-tandem mass spectrometry, and (1)H, (13)C, and two-dimensional NMR. This new chromophore exhibited a UV absorbance maximum at 343 nm and fluorescence at 410 nm when excited at 343 nm. Analysis of the purified compound by reversed-phase HPLC with in-line electrospray ionization mass spectrometry revealed a molecular mass of 370 Da. One- and two-dimensional NMR analyses elucidated the structure to be 1-(5-amino-5-carboxypentyl)-4-(5-amino-5-carboxypentylamino)-3-hydroxy-2,3-dihydropyridinium, a cross-link between the epsilon-amino groups of two lysine residues, and a five-carbon ring. Because this cross-link contains two lysine residues and a dihydropyridinium ring, we assigned it the trivial name of K2P. Quantitative determinations of K2P in individual normal human lens or cataract lens water-soluble and water-insoluble protein digests were made using a high-performance liquid chromatograph equipped with a diode array detector. These measurements revealed a significant enhancement of K2P in cataract lens proteins (613 +/- 362 pmol/mg of water-insoluble sonicate supernatant (WISS) protein or 85 +/- 51 pmol/mg of WS protein) when compared with aged normal human lens proteins (261 +/- 93 pmol/mg of WISS protein or 23 +/- 15 pmol/mg of water-soluble (WS) protein). These data provide chemical evidence for increased protein cross-linking during aging and cataract development in vivo. This new cross-link may serve as a quantitatively more significant biomarker for assessing the role of lens protein modifications during aging and in the pathogenesis of cataract.
Sigma-Aldrich
Proteinase K from Tritirachium album, lyophilized powder, ≥30 units/mg protein
Pricing and availability is not currently available.