MilliporeSigma
  • Analysis of repaglinide enantiomers in pharmaceutical formulations by capillary electrophoresis using 2,6-di-o-methyl-β-cyclodextrin as a chiral selector.
Analysis of repaglinide enantiomers in pharmaceutical formulations by capillary electrophoresis using 2,6-di-o-methyl-β-cyclodextrin as a chiral selector.
Journal of chromatographic science (2012-05-24)
Cen Li, Ye Jiang
ABSTRACT
This study used the general applicability of 2,6-didi-o-methyl-β-cyclodextrin (DM-β-CD) as the chiral selector in capillary electrophoresis for fast and efficient chiral separation of repaglinide enantiomers. A systematic study of the parameters affecting separation was performed with UV detection at 243 nm. The optimum conditions were determined to be 1.25% (w/v) DM-β-CD in 20 mM sodium phosphate (pH 2.5) as the running buffer and separation voltage at 20 kV. DM-β-CD had the best enantiomer resolution properties under the tested conditions, whereas other β-cyclodextrins showed inferior performances or no performance. The proposed method had a linear calibration curve in the concentration range of 12.5-400 µg/mL. The limit of detection was 100 ng/mL. The intra-day and inter-day precisions were 2.8 and 3.2%, respectively. Recoveries of 97.9-100.9% were obtained. The proposed method was fast and convenient, and was determined to be efficient for separating enantiomers and applicable for analyzing repaglinide enantiomers in quality control of pharmaceutical production.
Product Description
Repaglinide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methyl-β-cyclodextrin, average Mn 1310
Sigma-Aldrich
Repaglinide, ≥98% (HPLC), solid
Repaglinide for system suitability, European Pharmacopoeia (EP) Reference Standard