MilliporeSigma
  • Effect of temperature on the degree of conversion and working time of dual-cured resin cements exposed to different curing conditions.
Effect of temperature on the degree of conversion and working time of dual-cured resin cements exposed to different curing conditions.
Operative dentistry (2012-07-24)
M Oliveira, P F Cesar, M Giannini, F A Rueggeberg, J Rodrigues, C A Arrais
ABSTRACT
This study evaluated the degree of conversion (DC) and working time (WT) of two commercial, dual-cured resin cements polymerized at varying temperatures and under different curing-light accessible conditions, using Fourier transformed infrared analysis (FTIR). Calibra (Cal; Dentsply Caulk) and Variolink II (Ivoclar Vivadent) were tested at 25°C or preheated to 37°C or 50°C and applied to a similar-temperature surface of a horizontal attenuated-total-reflectance unit (ATR) attached to an infrared spectrometer. The products were polymerized using one of four conditions: direct light exposure only (600 mW/cm(2)) through a glass slide or through a 1.5- or 3.0-mm-thick ceramic disc (A2 shade, IPS e.max, Ivoclar Vivadent) or allowed to self-cure in the absence of light curing. FTIR spectra were recorded for 20 min (1 spectrum/s, 16 scans/spectrum, resolution 4 cm(-1)) immediately after application to the ATR. DC was calculated using standard techniques of observing changes in aliphatic-to-aromatic peak ratios precuring and 20-min postcuring as well as during each 1-second interval. Time-based monomer conversion analysis was used to determine WT at each temperature. DC and WT data (n=6) were analyzed by two-way analysis of variance and Tukey post hoc test (p=0.05). Higher temperatures increased DC regardless of curing mode and product. For Calibra, only the 3-mm-thick ceramic group showed lower DC than the other groups at 25°C (p=0.01830), while no significant difference was observed among groups at 37°C and 50°C. For Variolink, the 3-mm-thick ceramic group showed lower DC than the 1-mm-thick group only at 25°C, while the self-cure group showed lower DC than the others at all temperatures (p=0.00001). WT decreased with increasing temperature: at 37°C near 70% reduction and at 50°C near 90% for both products, with WT reduction reaching clinically inappropriate times in some cases (p=0.00001). Elevated temperature during polymerization of dual-cured cements increased DC. WT was reduced with elevated temperature, but the extent of reduction might not be clinically acceptable.
Sigma-Aldrich
Triethylene glycol dimethacrylate, contains 80-120 ppm MEHQ as inhibitor, 95%
Sigma-Aldrich
Triethylene glycol dimethacrylate, contains 200 ppm monomethyl ether hydroquinone as inhibitor, 99%
Sigma-Aldrich
Ytterbium(III) fluoride, anhydrous, powder, 99.98% trace metals basis