MilliporeSigma
  • Adiponectin blocks interleukin-18-mediated endothelial cell death via APPL1-dependent AMP-activated protein kinase (AMPK) activation and IKK/NF-kappaB/PTEN suppression.
Adiponectin blocks interleukin-18-mediated endothelial cell death via APPL1-dependent AMP-activated protein kinase (AMPK) activation and IKK/NF-kappaB/PTEN suppression.
The Journal of biological chemistry (2008-07-18)
Bysani Chandrasekar, William H Boylston, Kaliyamurthi Venkatachalam, Nicholas J G Webster, Sumanth D Prabhu, Anthony J Valente
ABSTRACT
The adipocyte-derived cytokine adiponectin is known to exert anti-inflammatory and anti-apoptotic effects. In patients with atherosclerotic cardiovascular disease, circulating levels of adiponectin correlate inversely with those of the proinflammatory, proapoptotic cytokine interleukin (IL)-18. The opposing actions of IL-18 and adiponectin on both cell survival and inflammation led us to investigate whether adiponectin signaling antagonizes IL-18-mediated endothelial cell death and to identify the underlying molecular mechanisms. Treatment with IL-18 suppressed Akt phosphorylation and its associated kinase activity, induced IkappaB kinase (IKK)-NF-kappaB-dependent PTEN activation, and promoted endothelial cell death. Pretreatment with adiponectin stimulated APPL1-dependent AMPK activation, reversed Akt inhibition in a phosphatidylinositol 3-kinase-dependent manner, blocked IKK-NF-kappaB-PTEN signaling, reduced caspase-3 activity, blocked Bax translocation, and inhibited endothelial cell death. The cytoprotective effect of adiponectin signaling was recapitulated by treatment with the pharmacological AMPK activator 5-aminoimidazole-4-carboxamide-1-beta-riboside. Collectively, these results demonstrated that adiponectin reverses IL-18-mediated endothelial cell death through an AMPK-associated mechanism, which may thus have therapeutic potential for diminishing IL-18-dependent vascular injury and inflammation.
Product Description
Roche
Cell Death Detection ELISAPLUS, sufficient for 96 multiwell tests (11774425001), sufficient for 10 x 96 multiwell tests (11920685001)
Pricing and availability is not currently available.