Merck
  • Disruption of the DT diaphorase (NQO1) gene in mice leads to increased menadione toxicity.
Disruption of the DT diaphorase (NQO1) gene in mice leads to increased menadione toxicity.
The Journal of biological chemistry (1998-04-29)
V Radjendirane, P Joseph, Y H Lee, S Kimura, A J Klein-Szanto, F J Gonzalez, A K Jaiswal
ABSTRACT
NAD(P)H:quinone oxidoreductase 1 (NQO1) is a flavoenzyme that catalyzes two-electron reductive metabolism and detoxification of quinones and their derivatives leading to protection of cells against redox cycling and oxidative stress. To examine the in vivo role of NQO1, a NQO1-null mouse was produced using targeted gene disruption. Mice lacking NQO1 gene expression showed no detectable phenotype and were indistinguishable from wild-type mice. However, NQO1-null mice exhibited increased toxicity when administered menadione compared with wild-type mice. These results establish a role for NQO1 in protection against quinone toxicity. The NQO1-null mice are a model for NQO1 deficiency in humans and can be used to determine the role of this enzyme in sensitivity to toxicity and carcinogenesis.