Merck
99100089
Opentrons Flex Thermocycler Gaskets
new
Opentrons Flex Thermocycler Gaskets
Select a Size
Pricing and availability is not currently available.
Skip To
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
EPDM rubber
manufacturer/tradename
991-00089
packaging
pkg of 10 count
compatibility
OPENTRONSFLEX™ Thermocycler Module GEN2
General description
OPENTRONSFLEX Thermocycler Gaskets are adhesive-backed Ethylene Propylene Diene Monomer (EPDM) seals designed for use with the fully-automated OPENTRONSFLEX Thermocycler. They come in a package of 10 seals. These seals need to be applied to the hood of the Thermocycler and can be reused up to 20 times. OPENTRONSFLEX Thermocycler Gaskets need to be sterilized before each use.
Legal Information
OPENTRONSFLEX is a trademark of Opentrons Labworks, Inc.