About This Item
UNSPSC Code:
41121608
Volume:
50 μL
Sterility:
sterile
Feature:
DNA free (Human)
PCR clean
RNase and DNase free
endotoxin free
pyrogen free
PCR clean
RNase and DNase free
endotoxin free
pyrogen free
Packaging:
pkg of 20 packs of 96 each
Manufacturer/tradename:
991-00101
Skip To
material
polypropylene
sterility
sterile
feature
DNA free (Human)
PCR clean
RNase and DNase free
endotoxin free
pyrogen free
manufacturer/tradename
991-00101
packaging
pkg of 20 packs of 96 each
volume
50 μL
Related Categories
General description
50 µL pipette tips for the Opentrons Flex, provided sterile in racks or blister packs (refills). Tips are clear, non-conducting polypropylene tips, free of human DNA, PCR inhibitors, RNase/DNase, pyrogens, and endotoxins. Opentrons Flex Tips and Racks are autoclavable.
Legal Information
AAW is a trademark of Millipore Corporation
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Opentrons is a registered trademark of Opentrons Labworks, Inc.