Skip To
packaging
pack of 1 ea
manufacturer/tradename
999-00204
technique(s)
analytical sample preparation: suitable
application(s)
clinical testing
compatibility
AAW™ Workstation & Flex Robot
Related Categories
General description
OPENTRONSFLEX™ Magnetic Block is a 96-well magnetic well plate holder containing static ring magnets, specifically designed for use with the AAW™ Workstation. Use it with the AAW™ Flex Gripper and skirted 96-well PCR or deep-well plates to facilitate quick and efficient separation using magnetic beads.
Legal Information
AAW is a trademark of Millipore Corporation
OPENTRONSFLEX is a trademark of Opentrons Labworks, Inc.