Merck
99900227
Opentrons Flex Waste Chute
new
Opentrons Flex Waste Chute
Slide 1 of 1
Select a Size
Pricing and availability is not currently available.
Skip To
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
packaging
pack of 1 ea
manufacturer/tradename
999-00227
technique(s)
analytical sample preparation: suitable
application(s)
waste management
compatibility
OPENTRONSFLEX™ Deck Expansion
General description
OPENTRONSFLEX Waste Chute occupies position D3 and is used to accept waste such as tips, tip racks, and plates in high volumes. The gripper will pick up the consumable and dispose it into the chute based on your workflow needs. The waste chute is neccesary for all high-throughput workflows and can be paired with the OPENTRONSFLEX Deck Expansion to maximize the throughput capabilities of the AAW Automated Assay Workstation.
Legal Information
AAW is a trademark of Millipore Corporation
OPENTRONSFLEX is a trademark of Opentrons Labworks, Inc.