Merck
99900244
AAW Flex Gripper
new
AAW Gripper
Select a Size
Pricing and availability is not currently available.
About This Item
UNSPSC Code:
41121521
Skip To
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
packaging
pack of 1 ea
manufacturer/tradename
999-00244
technique(s)
analytical sample preparation: suitable
application(s)
clinical testing
pharmaceutical
compatibility
Opentrons Flex Deck Expansion Slots
General description
The AAW Flex Gripper easily attaches to the gantry of the AAW Automated Assay Worksatation, allowing for efficient movement of labware across the deck and onto modules like the Magnetic Block or Heater-Shaker. It also provides access to additional side slots on the main deck for replenishing tips or labware. Compatible with any pipette configuration, this versatile gripper enhances the flexibility of your on-deck workflow.Included with the gripper is a calibration pin that facilitates automatic calibration, ensuring precise positioning.
Legal Information
AAW is a trademark of Millipore Corporation