Skip to Content
Merck

99900256

AAW Automated Assay Workstation - Assay Ready

new

AAW Automated Assay Workstation - Assay Ready

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

UNSPSC Code:
41121501
NACRES:
NC.01
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

packaging

pack of 1 ea

manufacturer/tradename

999-00256

technique(s)

activity assay: suitable

suitability

suitable for immunoassay
suitable for molecular biology

application(s)

recombinant protein

compatibility

Opentrons® Heater-Shaker Module
Opentrons® Magnetic Block
Opentrons® Temperature Module
Opentrons® Thermocycler Module
HEPA (High Efficiency Particulate Air) module

General description

The AAW Automated Assay Workstation - Assay Ready is a robotic platform for the life science industry designed for automating assays and sample preparation for a broad range of applications. Thanks to the power of Opentrons(R), it comes with a variety of modular hardware accessories that accomodate unit operations including pipetting, heating, shaking, magnetic resin pulldowns and more. It also comes with full access to the Millipore Assay Protocol Library. This is a library of automation protocols using common Millipore and Sigma-Aldrich assay kits and reagents that can be downloaded and are verified to perform consistently.

Application

Automation of assays and sample preparations for laboratory research and discovery, including immunoassays, protein purification, proteomics, molecular biology, and cell biology.

Features and Benefits

  • Gripper - for moving labware around on deck
  • Single and multi-channel liquid handling
  • Magnetic block for magnetic resin purification steps
  • Heating and shaking from ambient to 95°C
  • Access to Millipore Assay Protocol Library
  • Modular, plug-and-play accessories sold allow you to easily flex to new applications in the future.

Legal Information

AAW is a trademark of Millipore Corporation
Opentrons is a registered trademark of Opentrons Labworks, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service