Merck
All Photos(3)

261319

Sigma-Aldrich

Yttrium

ingot, 99.9% trace rare earth metals basis

Empirical Formula (Hill Notation):
Y
CAS Number:
Molecular Weight:
88.91
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.23

Quality Level

assay

99.9% trace rare earth metals basis

form

ingot

resistivity

57 μΩ-cm, 20°C

bp

3338 °C (lit.)

mp

1522 °C (lit.)

density

4.469 g/mL at 25 °C (lit.)

SMILES string

[Y]

InChI

1S/Y

InChI key

VWQVUPCCIRVNHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service