Merck
All Photos(2)

267406

Sigma-Aldrich

Ruthenium

sponge, −100 mesh, 99.9% trace metals basis

Synonym(s):
Ruthenium black, Ruthenium element
Empirical Formula (Hill Notation):
Ru
CAS Number:
Molecular Weight:
101.07
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.23

Quality Level

assay

99.9% trace metals basis

form

sponge

resistivity

7.1 μΩ-cm, 0°C

particle size

−100 mesh

bp

3900 °C (lit.)

mp

2310 °C (lit.)

density

12.45 g/cm3 (lit.)

SMILES string

[Ru]

InChI

1S/Ru

InChI key

KJTLSVCANCCWHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Rafael E Rodríguez-Lugo et al.
Nature chemistry, 5(4), 342-347 (2013-03-21)
The development of an efficient catalytic process that mimics the enzymatic function of alcohol dehydrogenase is critical for using biomass alcohols for both the production of H2 as a chemical energy carrier and fine chemicals under waste-free conditions. Dehydrogenation of
Direct synthesis of pyrroles by dehydrogenative coupling of β-aminoalcohols with secondary alcohols catalyzed by ruthenium pincer complexes.
Dipankar Srimani et al.
Angewandte Chemie (International ed. in English), 52(14), 4012-4015 (2013-03-08)
Jérôme Le Nôtre et al.
ChemSusChem, 6(4), 693-700 (2013-03-05)
A highly efficient procedure for obtaining resin-grade isoidide through catalytic epimerization of isosorbide using a ruthenium-on-carbon (Ru/C) catalyst is reported. A comprehensive reaction-parameter variation study involving substrate concentration, catalyst (type of metal, support, and loading), initial pH value, hydrogen pressure
Mette T Petersen et al.
Organic letters, 15(8), 1986-1989 (2013-04-05)
A series of 5-substituted 2-pyrrolidinones was synthesized through a one-pot ruthenium alkylidene-catalyzed tandem RCM/isomerization/nucleophilic addition sequence. The intermediates resulting from RCM/isomerization showed reactivity toward electrophiles in aldol condensation reactions which provided a new entry for the total synthesis of the
M Beye et al.
Physical review letters, 110(18), 186101-186101 (2013-05-21)
We have studied the femtosecond dynamics following optical laser excitation of CO adsorbed on a Ru surface by monitoring changes in the occupied and unoccupied electronic structure using ultrafast soft x-ray absorption and emission. We recently reported [M. Dell'Angela et

Articles

Materials for Clean H2 Production from Bioethanol Reforming

Hydrogen is one of the most important resources in providing food, fuel, and chemical products for our everyday life. Sustainable catalytic hydrogen production from bioethanol has gained significant attention in recent years due to globally diminishing fossil fuel supplies, which have necessitated the search for new chemical feedstocks.

Related Content

Responsive Rare-Earth Materials and their Applications

The unique properties of the rare-earth elements and their alloys have brought them from relative obscurity to high profile use in common hightech applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service