Merck
All Photos(1)

611395

Supelco

Urea-15N2 solution

NMR reference standard, 0.1 M in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.

Empirical Formula (Hill Notation):
CH415N2O
Molecular Weight:
62.04
EC Number:

Quality Level

grade

NMR reference standard
analytical standard

isotopic purity

99 atom % 15N

concentration

0.1 M in DMSO-d6 (99.9 atom % D)

technique(s)

NMR: suitable

NMR tube size

5 mm × 8 in.

format

single component solution

Related Categories

Features and Benefits

Indirect detection

Quantity

5 mm O.D. tube contains 0.700 mL.
Each pack contains 10 tubes.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service