Merck
All Photos(1)

678139

Lithium bromide solution

1.5 M in THF

Linear Formula:
LiBr
CAS Number:
Molecular Weight:
86.85
MDL number:
PubChem Substance ID:
NACRES:
NA.21

concentration

1.5 M in THF

density

0.995 g/mL at 20 °C

SMILES string

[Li+].[Br-]

InChI

1S/BrH.Li/h1H;/q;+1/p-1

InChI key

AMXOYNBUYSYVKV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Mohammad M Mojtahedi et al.
Organic letters, 9(15), 2791-2793 (2007-06-22)
A room temperature convenient disproportionation or reduction of aldehydes prompted by lithium bromide and triethylamine is described in a solvent-free environment. Distribution of the products to selectively direct the process toward Cannizzaro or Tishchenko reactions is controlled by the type
Ngoc-Ly Hoang et al.
Journal of chromatography. A, 1205(1-2), 60-70 (2008-08-30)
The structure of starch molecules is relevant to nutrition and industrial applications. Size-exclusion chromatography (SEC, also known as GPC) of native starch generally suffers non-satisfactory repeatability and reproducibility of the dissolution and separation. This work combines two polar organic solvents:
R Quaderer et al.
Organic letters, 3(20), 3181-3184 (2001-09-28)
[reaction: see text] The alkanesulfonamide "safety-catch" resin has proven useful for Fmoc-based synthesis of C-terminal peptide thioesters. We now report that the yield of isolated thioester can increase significantly when the cleavage reaction is carried out in 2 M LiBr/THF
Zhongli Cai et al.
Radiation research, 159(3), 411-419 (2003-02-26)
An investigation of electron and hole transfer to oxidized guanine bases in DNA is reported. Guanine in DNA was preferentially oxidized by Br(2)(*-) at 298 K to 8-oxo-7,8-dihydro-guanine (8-oxo-G) and higher oxidation products. HPLC-EC analysis of irradiated DNA shows that
Sophie Nocquet-Thibault et al.
Organic letters, 15(8), 1842-1845 (2013-04-02)
Using (diacetoxyiodo)benzene in conjunction with simple bromide salts in ethanol allows the regioselective ethoxybromination of a wide range of enamides, thus yielding highly versatile α-bromo hemiaminals, which can then be engaged in a broad array of transformations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service