BR122765
BRAND® Bottle-Top Filter
NACRES:
NB.24
material
polyethersulfone membrane
sterility
sterile
packaging
pack of 12
manufacturer/tradename
BRAND 122765
pore size
0.45 μm
Related Categories
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Certificate of Analysis
Certificate of Origin