All Photos(2)

GF44272537

Iron

foil, thickness 0.9 mm, 25 mm diameter, purity 99.5%

Synonym(s):
FE000414
Linear Formula:
Fe
CAS Number:
Molecular Weight:
55.85
MDL number:
PubChem Substance ID:
NACRES:
NA.23

assay

99.5%

form

foil

manufacturer/tradename

Goodfellow 442-725-37

resistivity

9.71 μΩ-cm

diam.

25 mm

thickness

0.9 mm

diameter

25 mm

bp

2750 °C (lit.)

mp

1535 °C (lit.)

density

7.86 g/mL at 25 °C (lit.)

SMILES string

[Fe]

InChI

1S/Fe

InChI key

XEEYBQQBJWHFJM-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service