GF46722089

Iron

foil, thickness 1.63 mm, size 150 × 500 mm, purity 99.5%

Synonym(s):
FE000560
Linear Formula:
Fe
CAS Number:
Molecular Weight:
55.85
MDL number:
PubChem Substance ID:

assay

99.5%

form

foil

manufacturer/tradename

Goodfellow 467-220-89

resistivity

9.71 μΩ-cm

size

150 × 500 mm

thickness

1.63 mm

bp

2750 °C (lit.)

mp

1535 °C (lit.)

density

7.86 g/mL at 25 °C (lit.)

SMILES string

[Fe]

InChI

1S/Fe

InChI key

XEEYBQQBJWHFJM-UHFFFAOYSA-N

Related Categories

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

storage_class_code

11 - Combustible Solids

WGK Germany

nwg

Flash Point F

Not applicable

Flash Point C

Not applicable

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service