Merck
All Photos(2)

Documents

GF67418062

Iron

tube, 500mm, outside diameter 12.0mm, inside diameter 9mm, wall thickness 1.5mm, annealed, 99.5%

Sign Into View Organizational & Contract Pricing

Synonym(s):
Iron, FE007680
Linear Formula:
Fe
CAS Number:
Molecular Weight:
55.85
MDL number:
PubChem Substance ID:
NACRES:
NA.23

Assay

99.5%

form

tubes

manufacturer/tradename

Goodfellow 674-180-62

resistivity

9.71 μΩ-cm

L × wall thickness

500 mm × 1.5 mm

O.D. × I.D.

12.0 mm × 9.0 mm

bp

2750 °C (lit.)

mp

1535 °C (lit.)

density

7.86 g/mL at 25 °C (lit.)

SMILES string

[Fe]

InChI

1S/Fe

InChI key

XEEYBQQBJWHFJM-UHFFFAOYSA-N

Related Categories

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF34939345GF61545674GF73600512
Iron tube, 500mm, outside diameter 12.0mm, inside diameter 9mm, wall thickness 1.5mm, annealed, 99.5%

GF67418062

Iron

Iron tube, 200mm, outside diameter 12.0mm, inside diameter 9mm, wall thickness 1.5mm, annealed, 99.5%

GF34939345

Iron

Iron tube, 100mm, outside diameter 8.0mm, inside diameter 5mm, wall thickness 1.5mm, annealed, 99.5%

GF61545674

Iron

Iron tube, 1000mm, outside diameter 12.7mm, inside diameter 9.5mm, wall thickness 1.6mm, annealed, 99.5%

GF73600512

Iron

form

tubes

form

tubes

form

tubes

form

tubes

manufacturer/tradename

Goodfellow 674-180-62

manufacturer/tradename

Goodfellow 349-393-45

manufacturer/tradename

Goodfellow 615-456-74

manufacturer/tradename

Goodfellow 736-005-12

resistivity

9.71 μΩ-cm

resistivity

9.71 μΩ-cm

resistivity

9.71 μΩ-cm

resistivity

9.71 μΩ-cm

bp

2750 °C (lit.)

bp

2750 °C (lit.)

bp

2750 °C (lit.)

bp

2750 °C (lit.)

mp

1535 °C (lit.)

mp

1535 °C (lit.)

mp

1535 °C (lit.)

mp

1535 °C (lit.)

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Thomas A Russo et al.
Infection and immunity, 82(6), 2356-2367 (2014-03-26)
Hypervirulent (hypermucoviscous) Klebsiella pneumoniae (hvKP) strains are an emerging variant of "classical" K. pneumoniae (cKP) that cause organ and life-threatening infection in healthy individuals. An understanding of hvKP-specific virulence mechanisms that enabled evolution from cKP is limited. Observations by our
Laura M van Staalduinen et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(14), 5171-5176 (2014-04-08)
The enzymes PhnY and PhnZ comprise an oxidative catabolic pathway that enables marine bacteria to use 2-aminoethylphosphonic acid as a source of inorganic phosphate. PhnZ is notable for catalyzing the oxidative cleavage of a carbon-phosphorus bond using Fe(II) and dioxygen
50 years ago in the Journal of Pediatrics: Iron metabolism in premature infants: II. Prevention of iron deficiency.
Pamela J Kling
The Journal of pediatrics, 164(4), 768-768 (2014-03-22)
Joseph D Mancias et al.
Nature, 509(7498), 105-109 (2014-04-04)
Autophagy, the process by which proteins and organelles are sequestered in double-membrane structures called autophagosomes and delivered to lysosomes for degradation, is critical in diseases such as cancer and neurodegeneration. Much of our understanding of this process has emerged from
Chenyan Lv et al.
Biochemistry, 53(14), 2232-2241 (2014-04-01)
From an evolutionary point of view, plant and animal ferritins arose from a common ancestor, but plant ferritin exhibits different features as compared with the animal analogue. One major difference is that the 4-fold channels naturally occurring in plant ferritin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service