Merck
All Photos(2)

GF75379977

Platinum

tube, 1000mm, outside diameter 0.4mm, inside diameter 0.1mm, wall thickness 0.15mm, as drawn, 99.95%

Synonym(s):
Platinum, PT007100
Empirical Formula (Hill Notation):
Pt
CAS Number:
Molecular Weight:
195.08
MDL number:
PubChem Substance ID:
NACRES:
NA.23

assay

99.95%

form

tubes

manufacturer/tradename

Goodfellow 753-799-77

resistivity

10.6 μΩ-cm, 20°C

L × thickness

1000 mm × 0.15 mm

O.D. × I.D.

0.4 mm × 0.1 mm

bp

3827 °C (lit.)

mp

1772 °C (lit.)

density

21.45 g/cm3 (lit.)

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

Related Categories

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service