GF93054865

Samarium

foil, 25x25mm, thickness 0.25mm, as rolled, 99%

Synonym(s):
SM000230, Samarium
Empirical Formula (Hill Notation):
Sm
CAS Number:
Molecular Weight:
150.36
MDL number:
PubChem Substance ID:
NACRES:
NA.23

assay

≥99%

form

foil

manufacturer/tradename

Goodfellow 930-548-65

resistivity

91.4 μΩ-cm, 0°C

bp

1794 °C (lit.)

mp

1074 °C (lit.)

density

7.47 g/mL at 25 °C (lit.)

SMILES string

[Sm]

InChI

1S/Sm

InChI key

KZUNJOHGWZRPMI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

pictograms

Flame

signalword

Warning

hcodes

hazcat

Water-react. 3

storage_class_code

4.3 - Hazardous materials, which set free flammable gases upon contact with water

WGK Germany

WGK 3

Flash Point F

Not applicable

Flash Point C

Not applicable

Certificate of Analysis

Certificate of Origin

A N Serafini
Cancer, 88(12 Suppl), 2934-2939 (2000-07-18)
In patients with bone pain due to metastatic disease, intravenous systemic radioisotope therapy may be a useful adjunct to other methods for palliating pain. Various studies have been performed utilizing a short-lived radioisotope conjugated to a tetraphosphonate (samarium 153 lexidronam)...
K C Nicolaou et al.
Angewandte Chemie (International ed. in English), 48(39), 7140-7165 (2009-08-29)
Introduced by Henri Kagan more than three decades ago, samarium diiodide (SmI(2)) has found increasing application in chemical synthesis. This single-electron reducing agent has been particularly useful in C-C bond formations, including those found in total synthesis endeavors. This Review...
Hiroshi Suizu et al.
Organic letters, 17(1), 126-129 (2014-12-17)
A new synthetic route to clavilactone B, a naturally occurring inhibitor of epidermal growth factor receptor (EGFR) tyrosine kinase, is disclosed. The route features a sequential samarium-mediated radical cyclization-fragmentation of an indanone derivative, which provides rapid access to a 10-membered...
José M Concellón et al.
Chemical Society reviews, 39(11), 4103-4113 (2010-08-05)
Samarium metal and samarium diiodide have become important tools as selective cyclopropanating agents in organic synthesis due to their high chemo- and stereoselectivity. Therefore, Sm and SmI(2) are the ideal reagents to prepare cyclopropane derivatives. This tutorial review highlights C-C...
Christine Beemelmanns et al.
Chemical Society reviews, 40(5), 2199-2210 (2011-01-19)
In this tutorial review we discuss recent advances in the field of ketyl-(het)arene cyclisations promoted by samarium diiodide and related processes. Couplings of samarium ketyls with carbon-carbon multiple bonds are perhaps the most useful reactions to create carbocycles and heterocycles...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service