Documents

GF94758991

Palladium

foil, light tested, 50x50mm, thickness 0.047mm, as rolled, 99.95%

Synonym(s):
PD000262, Palladium
Linear Formula:
Pd
CAS Number:
Molecular Weight:
106.42
MDL number:
PubChem Substance ID:
NACRES:
NA.23

assay

≥99.95%

form

foil

manufacturer/tradename

Goodfellow 947-589-91

resistivity

9.96 μΩ-cm, 20°C

bp

2970 °C (lit.)

mp

1554 °C (lit.)

density

12.02 g/cm3 (lit.)

SMILES string

[Pd]

InChI

1S/Pd

InChI key

KDLHZDBZIXYQEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

storage_class_code

13 - Non Combustible Solids

WGK Germany

nwg

Flash Point F

Not applicable

Flash Point C

Not applicable

Certificate of Analysis

Certificate of Origin

Simon De Corte et al.
Microbial biotechnology, 5(1), 5-17 (2011-05-11)
While precious metals are available to a very limited extent, there is an increasing demand to use them as catalyst. This is also true for palladium (Pd) catalysts and their sustainable recycling and production are required. Since Pd catalysts exist...
M Angeles Fernández-Ibañez et al.
Molecules (Basel, Switzerland), 18(9), 10108-10121 (2013-08-27)
The dual activation of simple substrates by the combination of organocatalysis and palladium catalysis has been successfully applied in a variety of different asymmetric transformations. Thus, the asymmetric a-allylation of carbonyl compounds, a-fluorination of acyl derivatives, decarboxylative protonation of β-dicarbonyl...
Mélanie Platon et al.
Chemical Society reviews, 41(10), 3929-3968 (2012-03-27)
A survey highlighting the most recent palladium catalytic systems produced and their performances for progress in direct synthesis of indole backbones by heterocarbocyclization of reactive substrates is provided. The discussion is developed in relation with the principles of sustainable chemistry...
Masahiro Yoshida
Chemical & pharmaceutical bulletin, 60(3), 285-299 (2012-03-03)
It is known that propargylic compounds having an ester and a halide at the propargylic positions react with palladium complexes leading to π-propargylpalladium and allenylpalladium complexes, which cause various transformations in the presence of the reactants. The aim of the...
Masahiro Toyota
Natural product communications, 8(7), 999-1004 (2013-08-29)
A novel palladium-catalyzed intramolecular oxidative alkylation of unactivated olefins is described. This protocol was devised to solve one of the drawbacks of the original palladium-catalyzed cycloalkenylation that we developed. We call this new procedure the 'second generation palladium-catalyzed cycloalkenylation'. This...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service